Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 65635808-65636081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP112
Entrez Gene ID
201134
Gene Name
centrosomal protein 112
Gene Aliases
CCDC46, MACOCO
UniGene
Hs.408676
Gene Chromosome Location
Chr.17: 65635540-66192131 on Build GRCh38
Transcripts
NM_001037325.2 NM_001199165.2 NM_001302891.1 NR_126542.1 XM_005257119.4 XM_005257125.3 XM_005257126.4 XM_006721740.2 XM_011524461.2 XM_011524464.2 XM_011524465.2 XM_011524466.2 XM_011524467.2 XM_017024302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
400536 c.2867A>T p.*956L Chr.17: 65635972-65635972 on Build GRCh38
400535 c.2867A>T p.*956L Chr.17: 65635972-65635972 on Build GRCh38
400537 c.635A>T p.*212L Chr.17: 65635972-65635972 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199771110 Chr.17: 65635951-65635951 on Build GRCh38
rs891363627 Chr.17: 65635944-65635944 on Build GRCh38
rs369663847 Chr.17: 65635939-65635939 on Build GRCh38
rs575808271 Chr.17: 65635998-65635998 on Build GRCh38
rs77414935 Chr.17: 65635956-65635956 on Build GRCh38
rs79781034 Chr.17: 65635990-65635990 on Build GRCh38
rs773072735 Chr.17: 65635960-65635960 on Build GRCh38
rs776688475 Chr.17: 65635952-65635952 on Build GRCh38
rs188564014 Chr.17: 65635966-65635966 on Build GRCh38
rs184186798 Chr.17: 65635847-65635847 on Build GRCh38
rs143768706 Chr.17: 65635872-65635872 on Build GRCh38
rs369671333 Chr.17: 65636041-65636041 on Build GRCh38
rs996100324 Chr.17: 65635864-65635864 on Build GRCh38
rs935603414 Chr.17: 65635908-65635908 on Build GRCh38
rs764718535 Chr.17: 65636012-65636012 on Build GRCh38
rs756550480 Chr.17: 65635999-65635999 on Build GRCh38
rs373721035 Chr.17: 65636016-65636018 on Build GRCh38
rs779064228 Chr.17: 65635933-65635933 on Build GRCh38
rs925289261 Chr.17: 65636030-65636030 on Build GRCh38
rs772780455 Chr.17: 65636015-65636017 on Build GRCh38
rs924910458 Chr.17: 65635877-65635877 on Build GRCh38
rs767059196 Chr.17: 65635994-65635994 on Build GRCh38
rs760021753 Chr.17: 65635940-65635940 on Build GRCh38
rs567048300 Chr.17: 65635965-65635965 on Build GRCh38
rs538533857 Chr.17: 65636049-65636049 on Build GRCh38

View Full Product Details