Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46648826-46649053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT3
Entrez Gene ID
147906
Gene Name
dishevelled binding antagonist of beta catenin 3
Gene Aliases
DAPPER3, RRR1
UniGene
Hs.515490
Gene Chromosome Location
Chr.19: 46647564-46661138 on Build GRCh38
Transcripts
NM_001301046.1 NM_145056.2 XM_011526499.2 XM_011526500.2 XM_017026334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3787728 c.1419C>T p.C473C Chr.19: 46648953-46648953 on Build GRCh38
5695767 c.1368G>A p.R456R Chr.19: 46649004-46649004 on Build GRCh38
5421677 c.788G>A p.R263H Chr.19: 46648909-46648909 on Build GRCh38
5692670 c.1321C>T p.P441S Chr.19: 46649051-46649051 on Build GRCh38
5770872 c.660C>T p.P220P Chr.19: 46649037-46649037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050705171 Chr.19: 46648883-46648883 on Build GRCh38
rs775908766 Chr.19: 46648955-46648955 on Build GRCh38
rs927652781 Chr.19: 46648851-46648851 on Build GRCh38
rs940338091 Chr.19: 46648854-46648854 on Build GRCh38
rs999489016 Chr.19: 46648999-46648999 on Build GRCh38
rs753977517 Chr.19: 46649007-46649007 on Build GRCh38
rs774957424 Chr.19: 46648885-46648885 on Build GRCh38
rs1035991464 Chr.19: 46648856-46648856 on Build GRCh38
rs1030931556 Chr.19: 46649018-46649018 on Build GRCh38
rs930353192 Chr.19: 46648872-46648872 on Build GRCh38
rs903068397 Chr.19: 46648925-46648925 on Build GRCh38
rs898926309 Chr.19: 46648868-46648868 on Build GRCh38
rs767969446 Chr.19: 46648943-46648943 on Build GRCh38
rs559668585 Chr.19: 46648861-46648861 on Build GRCh38
rs1009260109 Chr.19: 46648905-46648905 on Build GRCh38
rs760971337 Chr.19: 46648970-46648970 on Build GRCh38
rs1018856624 Chr.19: 46648919-46648919 on Build GRCh38
rs764169266 Chr.19: 46648990-46648990 on Build GRCh38
rs958133582 Chr.19: 46649027-46649027 on Build GRCh38
rs760016904 Chr.19: 46648900-46648900 on Build GRCh38
rs771489153 Chr.19: 46648871-46648871 on Build GRCh38

View Full Product Details