Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107945190-107945449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
UniGene
Hs.109087
Gene Chromosome Location
Chr.4: 107931549-107953461 on Build GRCh38
Transcripts
NM_183075.2 XM_005262717.2 XM_005262720.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
280266 c.923C>A p.S308Y Chr.4: 107945402-107945402 on Build GRCh38
4121212 c.888T>C p.S296S Chr.4: 107945367-107945367 on Build GRCh38
7363545 c.842T>A p.F281Y Chr.4: 107945321-107945321 on Build GRCh38
4121211 c.867A>C p.Q289H Chr.4: 107945346-107945346 on Build GRCh38
6639268 c.780A>C p.E260D Chr.4: 107945259-107945259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756893121 Chr.4: 107945335-107945335 on Build GRCh38
rs766883559 Chr.4: 107945304-107945304 on Build GRCh38
rs200611321 Chr.4: 107945408-107945408 on Build GRCh38
rs973278785 Chr.4: 107945351-107945351 on Build GRCh38
rs1027540127 Chr.4: 107945398-107945398 on Build GRCh38
rs942548642 Chr.4: 107945220-107945220 on Build GRCh38
rs760549002 Chr.4: 107945249-107945249 on Build GRCh38
rs373081734 Chr.4: 107945409-107945409 on Build GRCh38
rs773121382 Chr.4: 107945241-107945242 on Build GRCh38
rs556388568 Chr.4: 107945247-107945247 on Build GRCh38
rs764172871 Chr.4: 107945264-107945264 on Build GRCh38
rs764450466 Chr.4: 107945402-107945402 on Build GRCh38
rs377203075 Chr.4: 107945246-107945246 on Build GRCh38
rs200772412 Chr.4: 107945228-107945228 on Build GRCh38
rs61746386 Chr.4: 107945293-107945293 on Build GRCh38
rs752056740 Chr.4: 107945318-107945318 on Build GRCh38
rs748913829 Chr.4: 107945216-107945216 on Build GRCh38
rs1041397010 Chr.4: 107945326-107945326 on Build GRCh38
rs369211359 Chr.4: 107945294-107945294 on Build GRCh38
rs776577604 Chr.4: 107945275-107945275 on Build GRCh38
rs1052060667 Chr.4: 107945359-107945359 on Build GRCh38
rs1003330304 Chr.4: 107945331-107945331 on Build GRCh38
rs754089373 Chr.4: 107945419-107945419 on Build GRCh38
rs369116680 Chr.4: 107945367-107945367 on Build GRCh38
rs1020678112 Chr.4: 107945296-107945296 on Build GRCh38
rs758876011 Chr.4: 107945299-107945299 on Build GRCh38
rs148477072 Chr.4: 107945329-107945329 on Build GRCh38
rs762698300 Chr.4: 107945332-107945332 on Build GRCh38
rs397514515 Chr.4: 107945263-107945263 on Build GRCh38
rs755453405 Chr.4: 107945320-107945320 on Build GRCh38
rs746892717 Chr.4: 107945405-107945405 on Build GRCh38
rs774131003 Chr.4: 107945234-107945234 on Build GRCh38
rs777274185 Chr.4: 107945328-107945328 on Build GRCh38
rs761846487 Chr.4: 107945415-107945415 on Build GRCh38
rs762090399 Chr.4: 107945277-107945277 on Build GRCh38
rs745724197 Chr.4: 107945238-107945238 on Build GRCh38
rs761095930 Chr.4: 107945348-107945348 on Build GRCh38
rs777170829 Chr.4: 107945211-107945211 on Build GRCh38
rs779834468 Chr.4: 107945393-107945393 on Build GRCh38

View Full Product Details