Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119672132-119672377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG3
Entrez Gene ID
9531
Gene Name
BCL2 associated athanogene 3
Gene Aliases
BAG-3, BIS, CAIR-1, MFM6
UniGene
Hs.523309
Gene Chromosome Location
Chr.10: 119651347-119677819 on Build GRCh38
Transcripts
NM_004281.3 XM_005270287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6258254 c.589G>T p.G197C Chr.10: 119672336-119672336 on Build GRCh38
2055642 c.566C>T p.P189L Chr.10: 119672313-119672313 on Build GRCh38
1184363 c.614A>G p.Y205C Chr.10: 119672361-119672361 on Build GRCh38
4012090 c.508C>T p.R170W Chr.10: 119672255-119672255 on Build GRCh38
3434637 c.578G>A p.R193K Chr.10: 119672325-119672325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74157690 Chr.10: 119672353-119672353 on Build GRCh38
rs927441881 Chr.10: 119672329-119672329 on Build GRCh38
rs964476630 Chr.10: 119672175-119672175 on Build GRCh38
rs867901145 Chr.10: 119672248-119672248 on Build GRCh38
rs766766600 Chr.10: 119672304-119672304 on Build GRCh38
rs770048996 Chr.10: 119672207-119672207 on Build GRCh38
rs768201611 Chr.10: 119672347-119672347 on Build GRCh38
rs372060324 Chr.10: 119672172-119672172 on Build GRCh38
rs539400088 Chr.10: 119672205-119672205 on Build GRCh38
rs755529596 Chr.10: 119672237-119672237 on Build GRCh38
rs768270028 Chr.10: 119672263-119672263 on Build GRCh38
rs750426195 Chr.10: 119672336-119672336 on Build GRCh38
rs374243122 Chr.10: 119672229-119672229 on Build GRCh38
rs536339219 Chr.10: 119672207-119672211 on Build GRCh38
rs730880054 Chr.10: 119672301-119672301 on Build GRCh38
rs755969300 Chr.10: 119672337-119672337 on Build GRCh38
rs556000090 Chr.10: 119672352-119672352 on Build GRCh38
rs794728978 Chr.10: 119672259-119672259 on Build GRCh38
rs570390738 Chr.10: 119672188-119672188 on Build GRCh38
rs886046754 Chr.10: 119672311-119672311 on Build GRCh38
rs140904592 Chr.10: 119672256-119672256 on Build GRCh38
rs536117098 Chr.10: 119672331-119672331 on Build GRCh38
rs528288616 Chr.10: 119672330-119672330 on Build GRCh38
rs955856672 Chr.10: 119672216-119672216 on Build GRCh38
rs914284920 Chr.10: 119672232-119672232 on Build GRCh38
rs1023800067 Chr.10: 119672162-119672162 on Build GRCh38
rs780054019 Chr.10: 119672342-119672342 on Build GRCh38
rs376601713 Chr.10: 119672230-119672230 on Build GRCh38
rs772491154 Chr.10: 119672294-119672294 on Build GRCh38
rs753386051 Chr.10: 119672320-119672320 on Build GRCh38
rs550619187 Chr.10: 119672187-119672187 on Build GRCh38
rs762317370 Chr.10: 119672283-119672283 on Build GRCh38
rs982369319 Chr.10: 119672181-119672181 on Build GRCh38
rs763170019 Chr.10: 119672293-119672294 on Build GRCh38
rs796123466 Chr.10: 119672275-119672275 on Build GRCh38
rs112929734 Chr.10: 119672296-119672296 on Build GRCh38
rs749017720 Chr.10: 119672260-119672260 on Build GRCh38
rs1047671466 Chr.10: 119672354-119672354 on Build GRCh38
rs778924812 Chr.10: 119672220-119672220 on Build GRCh38
rs755015322 Chr.10: 119672207-119672207 on Build GRCh38
rs752722473 Chr.10: 119672325-119672325 on Build GRCh38
rs780973226 Chr.10: 119672239-119672239 on Build GRCh38
rs1051673569 Chr.10: 119672333-119672333 on Build GRCh38
rs1038821617 Chr.10: 119672349-119672349 on Build GRCh38
rs748893116 Chr.10: 119672344-119672344 on Build GRCh38
rs1017390122 Chr.10: 119672164-119672164 on Build GRCh38
rs773750158 Chr.10: 119672265-119672265 on Build GRCh38
rs978476877 Chr.10: 119672186-119672186 on Build GRCh38
rs928320768 Chr.10: 119672190-119672190 on Build GRCh38
rs201487919 Chr.10: 119672321-119672321 on Build GRCh38
rs200479768 Chr.10: 119672255-119672255 on Build GRCh38

View Full Product Details