Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97910318-97910563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMO
Entrez Gene ID
51531
Gene Name
tRNA methyltransferase O
Gene Aliases
C9orf156, HSPC219, NAP1
UniGene
Hs.9196
Gene Chromosome Location
Chr.9: 97876321-97922570 on Build GRCh38
Transcripts
NM_016481.4 XM_005252047.3 XM_005252048.4 XM_005252049.4 XM_005252050.4 XM_011518776.2 XM_011518777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5495529 c.606G>C p.Q202H Chr.9: 97910420-97910420 on Build GRCh38
5804726 c.593G>T p.C198F Chr.9: 97910433-97910433 on Build GRCh38
1624344 c.470A>T p.K157M Chr.9: 97910556-97910556 on Build GRCh38
7412812 c.676A>C p.T226P Chr.9: 97910350-97910350 on Build GRCh38
6476459 c.507G>T p.V169V Chr.9: 97910519-97910519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759787250 Chr.9: 97910381-97910381 on Build GRCh38
rs1017890055 Chr.9: 97910398-97910398 on Build GRCh38
rs537906847 Chr.9: 97910412-97910412 on Build GRCh38
rs922555506 Chr.9: 97910413-97910414 on Build GRCh38
rs754853403 Chr.9: 97910453-97910453 on Build GRCh38
rs772887956 Chr.9: 97910540-97910540 on Build GRCh38
rs374832074 Chr.9: 97910349-97910349 on Build GRCh38
rs567392067 Chr.9: 97910411-97910411 on Build GRCh38
rs765381652 Chr.9: 97910366-97910366 on Build GRCh38
rs749423892 Chr.9: 97910356-97910356 on Build GRCh38
rs768675466 Chr.9: 97910346-97910346 on Build GRCh38
rs746070951 Chr.9: 97910488-97910488 on Build GRCh38
rs781122406 Chr.9: 97910469-97910469 on Build GRCh38
rs143780090 Chr.9: 97910402-97910402 on Build GRCh38
rs201638443 Chr.9: 97910342-97910342 on Build GRCh38
rs770962621 Chr.9: 97910479-97910479 on Build GRCh38
rs977234325 Chr.9: 97910452-97910453 on Build GRCh38
rs750045705 Chr.9: 97910414-97910414 on Build GRCh38
rs772380406 Chr.9: 97910506-97910506 on Build GRCh38
rs999600654 Chr.9: 97910362-97910362 on Build GRCh38
rs574871568 Chr.9: 97910415-97910415 on Build GRCh38
rs571150996 Chr.9: 97910528-97910528 on Build GRCh38
rs983876205 Chr.9: 97910351-97910352 on Build GRCh38
rs775973178 Chr.9: 97910400-97910400 on Build GRCh38
rs199556128 Chr.9: 97910372-97910372 on Build GRCh38
rs772279326 Chr.9: 97910385-97910385 on Build GRCh38
rs777236790 Chr.9: 97910527-97910527 on Build GRCh38
rs778989810 Chr.9: 97910480-97910480 on Build GRCh38
rs370124786 Chr.9: 97910513-97910513 on Build GRCh38
rs777707868 Chr.9: 97910350-97910350 on Build GRCh38
rs925814803 Chr.9: 97910462-97910462 on Build GRCh38
rs774494137 Chr.9: 97910379-97910379 on Build GRCh38
rs751435259 Chr.9: 97910436-97910436 on Build GRCh38
rs768917047 Chr.9: 97910519-97910519 on Build GRCh38
rs187040706 Chr.9: 97910475-97910475 on Build GRCh38
rs143064742 Chr.9: 97910409-97910409 on Build GRCh38
rs373301254 Chr.9: 97910529-97910529 on Build GRCh38
rs945981440 Chr.9: 97910424-97910424 on Build GRCh38
rs779678919 Chr.9: 97910425-97910425 on Build GRCh38
rs761159317 Chr.9: 97910517-97910517 on Build GRCh38

View Full Product Details