Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 12678318-12678545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP76
Entrez Gene ID
79959
Gene Name
centrosomal protein 76
Gene Aliases
C18orf9, HsT1705
UniGene
Hs.236940
Gene Chromosome Location
Chr.18: 12661733-12702777 on Build GRCh38
Transcripts
NM_001271989.1 NM_024899.3 NR_073537.1 XM_005258149.4 XM_017025981.1 XM_017025982.1 XM_017025983.1 XM_017025984.1 XM_017025985.1 XM_017025986.1 XM_017025987.1 NM_147163.1

Gene Symbol
PSMG2
Entrez Gene ID
56984
Gene Name
proteasome assembly chaperone 2
Gene Aliases
CLAST3, HCCA3, HsT1707, MDS003, PAC2, TNFSF5IP1
UniGene
Hs.464652
Gene Chromosome Location
Chr.18: 12658739-12725740 on Build GRCh38
Transcripts
NM_001271989.1 NM_024899.3 NR_073537.1 XM_005258149.4 XM_017025981.1 XM_017025982.1 XM_017025983.1 XM_017025984.1 XM_017025985.1 XM_017025986.1 XM_017025987.1 NM_147163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5828648 c.1290-3delT p.? Chr.18: 12678445-12678445 on Build GRCh38
5006070 c.1290-10_1290-9insT p.? Chr.18: 12678451-12678452 on Build GRCh38
229590 c.1313C>T p.P438L Chr.18: 12678419-12678419 on Build GRCh38
4839042 c.1301A>C p.K434T Chr.18: 12678431-12678431 on Build GRCh38
4671707 c.1389G>A p.Q463Q Chr.18: 12678343-12678343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12966424 Chr.18: 12678487-12678487 on Build GRCh38
rs779390805 Chr.18: 12678391-12678391 on Build GRCh38
rs764987986 Chr.18: 12678460-12678460 on Build GRCh38
rs768627406 Chr.18: 12678419-12678419 on Build GRCh38
rs763149385 Chr.18: 12678476-12678482 on Build GRCh38
rs754179948 Chr.18: 12678371-12678371 on Build GRCh38
rs765985140 Chr.18: 12678488-12678488 on Build GRCh38
rs150533868 Chr.18: 12678504-12678504 on Build GRCh38
rs772552404 Chr.18: 12678459-12678459 on Build GRCh38
rs772924378 Chr.18: 12678365-12678365 on Build GRCh38
rs747134192 Chr.18: 12678407-12678407 on Build GRCh38
rs770177339 Chr.18: 12678444-12678445 on Build GRCh38
rs958174060 Chr.18: 12678343-12678343 on Build GRCh38
rs759114067 Chr.18: 12678411-12678411 on Build GRCh38
rs796927503 Chr.18: 12678516-12678516 on Build GRCh38
rs774297640 Chr.18: 12678426-12678426 on Build GRCh38
rs762832727 Chr.18: 12678415-12678415 on Build GRCh38
rs370679694 Chr.18: 12678347-12678347 on Build GRCh38
rs755375295 Chr.18: 12678377-12678377 on Build GRCh38
rs771036468 Chr.18: 12678408-12678408 on Build GRCh38
rs748498676 Chr.18: 12678366-12678366 on Build GRCh38
rs777672974 Chr.18: 12678405-12678405 on Build GRCh38
rs528253123 Chr.18: 12678454-12678454 on Build GRCh38
rs773563369 Chr.18: 12678457-12678457 on Build GRCh38
rs761592722 Chr.18: 12678447-12678447 on Build GRCh38
rs987357774 Chr.18: 12678485-12678485 on Build GRCh38
rs997130485 Chr.18: 12678401-12678401 on Build GRCh38
rs199846298 Chr.18: 12678397-12678397 on Build GRCh38
rs772635325 Chr.18: 12678400-12678400 on Build GRCh38
rs753554956 Chr.18: 12678492-12678492 on Build GRCh38

View Full Product Details