Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2538459-2538696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAT1
Entrez Gene ID
221927
Gene Name
BRCA1 associated ATM activator 1
Gene Aliases
BAAT1, C7orf27, RMFSL
UniGene
Hs.520623
Gene Chromosome Location
Chr.7: 2537810-2555758 on Build GRCh38
Transcripts
NM_152743.3 XM_005249643.4 XM_011515177.2 XM_011515178.1 XM_011515179.2 XM_011515181.2 XM_011515182.2 XM_011515183.1 XM_011515184.2 XM_011515186.2 XM_017011833.1 XM_017011834.1 XM_017011835.1 XM_017011836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198985 c.1954G>A p.A652T Chr.7: 2538581-2538581 on Build GRCh38
4666006 c.2019C>T p.C673C Chr.7: 2538516-2538516 on Build GRCh38
96915 c.1931_1932GA>AG p.R644Q Chr.7: 2538603-2538604 on Build GRCh38
4666007 c.1902C>T p.F634F Chr.7: 2538633-2538633 on Build GRCh38
6612474 c.1939G>A p.D647N Chr.7: 2538596-2538596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374879575 Chr.7: 2538604-2538604 on Build GRCh38
rs754828716 Chr.7: 2538605-2538605 on Build GRCh38
rs71531463 Chr.7: 2538603-2538604 on Build GRCh38
rs1047615442 Chr.7: 2538597-2538597 on Build GRCh38
rs372436637 Chr.7: 2538528-2538528 on Build GRCh38
rs61627394 Chr.7: 2538547-2538547 on Build GRCh38
rs371142628 Chr.7: 2538669-2538669 on Build GRCh38
rs369763195 Chr.7: 2538657-2538657 on Build GRCh38
rs4719553 Chr.7: 2538604-2538604 on Build GRCh38
rs145833100 Chr.7: 2538494-2538494 on Build GRCh38
rs996224277 Chr.7: 2538626-2538626 on Build GRCh38
rs371847594 Chr.7: 2538540-2538540 on Build GRCh38
rs777864683 Chr.7: 2538534-2538534 on Build GRCh38
rs750400556 Chr.7: 2538656-2538656 on Build GRCh38
rs560003607 Chr.7: 2538660-2538660 on Build GRCh38
rs749058877 Chr.7: 2538489-2538489 on Build GRCh38
rs373392231 Chr.7: 2538632-2538632 on Build GRCh38
rs775940127 Chr.7: 2538642-2538642 on Build GRCh38
rs761439588 Chr.7: 2538527-2538529 on Build GRCh38
rs35445563 Chr.7: 2538601-2538602 on Build GRCh38
rs200502048 Chr.7: 2538610-2538610 on Build GRCh38
rs778770405 Chr.7: 2538609-2538609 on Build GRCh38
rs77015302 Chr.7: 2538668-2538668 on Build GRCh38
rs752965856 Chr.7: 2538590-2538590 on Build GRCh38
rs757399456 Chr.7: 2538599-2538599 on Build GRCh38
rs770559678 Chr.7: 2538583-2538583 on Build GRCh38
rs761266451 Chr.7: 2538513-2538513 on Build GRCh38
rs771781859 Chr.7: 2538580-2538580 on Build GRCh38
rs141726264 Chr.7: 2538506-2538506 on Build GRCh38
rs758503229 Chr.7: 2538532-2538532 on Build GRCh38
rs767892335 Chr.7: 2538661-2538662 on Build GRCh38
rs546426836 Chr.7: 2538557-2538557 on Build GRCh38
rs760319281 Chr.7: 2538582-2538582 on Build GRCh38
rs745312463 Chr.7: 2538670-2538670 on Build GRCh38
rs894725088 Chr.7: 2538552-2538552 on Build GRCh38
rs938813337 Chr.7: 2538543-2538543 on Build GRCh38
rs765658318 Chr.7: 2538588-2538588 on Build GRCh38
rs865893083 Chr.7: 2538508-2538508 on Build GRCh38
rs764245593 Chr.7: 2538595-2538595 on Build GRCh38
rs968879693 Chr.7: 2538510-2538510 on Build GRCh38
rs772861328 Chr.7: 2538581-2538581 on Build GRCh38
rs376101375 Chr.7: 2538594-2538594 on Build GRCh38
rs138077616 Chr.7: 2538673-2538673 on Build GRCh38
rs1028833727 Chr.7: 2538515-2538515 on Build GRCh38
rs376186402 Chr.7: 2538507-2538507 on Build GRCh38
rs747027466 Chr.7: 2538539-2538539 on Build GRCh38
rs774038377 Chr.7: 2538505-2538506 on Build GRCh38
rs368232132 Chr.7: 2538531-2538531 on Build GRCh38
rs570576002 Chr.7: 2538529-2538529 on Build GRCh38
rs777235172 Chr.7: 2538584-2538584 on Build GRCh38
rs767664883 Chr.7: 2538655-2538655 on Build GRCh38
rs142346283 Chr.7: 2538651-2538651 on Build GRCh38
rs760122674 Chr.7: 2538631-2538631 on Build GRCh38
rs748335252 Chr.7: 2538558-2538558 on Build GRCh38
rs186448943 Chr.7: 2538563-2538563 on Build GRCh38
rs147119058 Chr.7: 2538526-2538526 on Build GRCh38
rs978884737 Chr.7: 2538511-2538511 on Build GRCh38
rs113030000 Chr.7: 2538613-2538613 on Build GRCh38
rs778520339 Chr.7: 2538573-2538573 on Build GRCh38
rs148297849 Chr.7: 2538674-2538674 on Build GRCh38
rs2917726 Chr.7: 2538612-2538612 on Build GRCh38
rs777688561 Chr.7: 2538664-2538664 on Build GRCh38
rs916113390 Chr.7: 2538663-2538663 on Build GRCh38
rs368449590 Chr.7: 2538527-2538527 on Build GRCh38
rs758308165 Chr.7: 2538662-2538662 on Build GRCh38
rs761970275 Chr.7: 2538654-2538654 on Build GRCh38
rs763369342 Chr.7: 2538643-2538643 on Build GRCh38
rs546403080 Chr.7: 2538659-2538659 on Build GRCh38
rs761345692 Chr.7: 2538500-2538500 on Build GRCh38
rs751770538 Chr.7: 2538596-2538596 on Build GRCh38
rs1018617325 Chr.7: 2538495-2538495 on Build GRCh38
rs4719552 Chr.7: 2538603-2538603 on Build GRCh38
rs1016199390 Chr.7: 2538624-2538624 on Build GRCh38
rs747863539 Chr.7: 2538577-2538577 on Build GRCh38
rs369737188 Chr.7: 2538618-2538618 on Build GRCh38
rs775271322 Chr.7: 2538483-2538483 on Build GRCh38
rs768326465 Chr.7: 2538493-2538493 on Build GRCh38
rs775956979 Chr.7: 2538629-2538629 on Build GRCh38
rs768540693 Chr.7: 2538568-2538568 on Build GRCh38

View Full Product Details