Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218238647-218238882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC2
Entrez Gene ID
10109
Gene Name
actin related protein 2/3 complex subunit 2
Gene Aliases
ARC34, PNAS-139, PRO2446, p34-Arc
UniGene
Hs.529303
Gene Chromosome Location
Chr.2: 218217094-218254348 on Build GRCh38
Transcripts
NM_005731.3 NM_152862.2 XM_011510471.1 XM_017003113.1 XM_017003114.1 XM_017003115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3577654 c.346T>C p.L116L Chr.2: 218238741-218238741 on Build GRCh38
6090559 c.308C>T p.P103L Chr.2: 218238703-218238703 on Build GRCh38
1326427 c.379A>G p.K127E Chr.2: 218238774-218238774 on Build GRCh38
7228857 c.455+7A>G p.? Chr.2: 218238857-218238857 on Build GRCh38
461168 c.372C>G p.V124V Chr.2: 218238767-218238767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865949510 Chr.2: 218238840-218238840 on Build GRCh38
rs1028736942 Chr.2: 218238792-218238792 on Build GRCh38
rs775360539 Chr.2: 218238783-218238783 on Build GRCh38
rs762858923 Chr.2: 218238797-218238797 on Build GRCh38
rs147649871 Chr.2: 218238802-218238802 on Build GRCh38
rs780413026 Chr.2: 218238677-218238677 on Build GRCh38
rs779118927 Chr.2: 218238704-218238704 on Build GRCh38
rs1803065 Chr.2: 218238714-218238714 on Build GRCh38
rs774147455 Chr.2: 218238811-218238811 on Build GRCh38
rs774270199 Chr.2: 218238732-218238732 on Build GRCh38
rs984436737 Chr.2: 218238822-218238822 on Build GRCh38
rs201701777 Chr.2: 218238778-218238778 on Build GRCh38
rs11551126 Chr.2: 218238763-218238763 on Build GRCh38
rs761202059 Chr.2: 218238836-218238836 on Build GRCh38
rs754230692 Chr.2: 218238695-218238695 on Build GRCh38
rs201975910 Chr.2: 218238740-218238740 on Build GRCh38
rs373428220 Chr.2: 218238702-218238702 on Build GRCh38
rs10169718 Chr.2: 218238857-218238857 on Build GRCh38
rs772187884 Chr.2: 218238748-218238748 on Build GRCh38
rs369075041 Chr.2: 218238800-218238800 on Build GRCh38
rs749920529 Chr.2: 218238826-218238826 on Build GRCh38
rs11551125 Chr.2: 218238723-218238723 on Build GRCh38
rs761601654 Chr.2: 218238812-218238812 on Build GRCh38
rs267599209 Chr.2: 218238718-218238718 on Build GRCh38
rs202051398 Chr.2: 218238818-218238818 on Build GRCh38
rs777765139 Chr.2: 218238759-218238759 on Build GRCh38
rs757883904 Chr.2: 218238733-218238733 on Build GRCh38
rs769653246 Chr.2: 218238779-218238779 on Build GRCh38

View Full Product Details