Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5204272-5204545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1C4
Entrez Gene ID
1109
Gene Name
aldo-keto reductase family 1, member C4
Gene Aliases
3-alpha-HSD, C11, CDR, CHDR, DD-4, DD4, HAKRA
UniGene
Hs.567245
Gene Chromosome Location
Chr.10: 5196835-5218947 on Build GRCh38
Transcripts
NM_001818.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6597980 c.309_310insA p.L106fs*7 Chr.10: 5204433-5204434 on Build GRCh38
4404635 c.316C>T p.L106F Chr.10: 5204440-5204440 on Build GRCh38
4394886 c.265T>G p.F89V Chr.10: 5204389-5204389 on Build GRCh38
5904919 c.369+6G>A p.? Chr.10: 5204499-5204499 on Build GRCh38
465722 c.268T>C p.F90L Chr.10: 5204392-5204392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782625417 Chr.10: 5204448-5204448 on Build GRCh38
rs782439273 Chr.10: 5204445-5204445 on Build GRCh38
rs753701215 Chr.10: 5204385-5204385 on Build GRCh38
rs111844367 Chr.10: 5204463-5204463 on Build GRCh38
rs781850610 Chr.10: 5204406-5204406 on Build GRCh38
rs967427615 Chr.10: 5204454-5204454 on Build GRCh38
rs782019921 Chr.10: 5204348-5204349 on Build GRCh38
rs782648087 Chr.10: 5204430-5204431 on Build GRCh38
rs782428027 Chr.10: 5204512-5204512 on Build GRCh38
rs782349269 Chr.10: 5204491-5204491 on Build GRCh38
rs781961888 Chr.10: 5204414-5204414 on Build GRCh38
rs373078506 Chr.10: 5204494-5204494 on Build GRCh38
rs782018320 Chr.10: 5204362-5204362 on Build GRCh38
rs782008432 Chr.10: 5204514-5204514 on Build GRCh38
rs374435340 Chr.10: 5204449-5204449 on Build GRCh38
rs549307097 Chr.10: 5204469-5204469 on Build GRCh38
rs782699066 Chr.10: 5204523-5204523 on Build GRCh38
rs781973294 Chr.10: 5204403-5204403 on Build GRCh38
rs367596754 Chr.10: 5204404-5204404 on Build GRCh38
rs782677864 Chr.10: 5204465-5204465 on Build GRCh38
rs138767311 Chr.10: 5204408-5204408 on Build GRCh38
rs1016830358 Chr.10: 5204298-5204298 on Build GRCh38
rs201035277 Chr.10: 5204482-5204482 on Build GRCh38
rs980735108 Chr.10: 5204350-5204350 on Build GRCh38
rs963600454 Chr.10: 5204333-5204333 on Build GRCh38
rs554928750 Chr.10: 5204421-5204421 on Build GRCh38
rs370748293 Chr.10: 5204329-5204329 on Build GRCh38
rs782237290 Chr.10: 5204490-5204490 on Build GRCh38
rs782423248 Chr.10: 5204358-5204358 on Build GRCh38
rs782216757 Chr.10: 5204463-5204464 on Build GRCh38
rs782789040 Chr.10: 5204410-5204410 on Build GRCh38
rs371830854 Chr.10: 5204438-5204438 on Build GRCh38
rs782452602 Chr.10: 5204478-5204478 on Build GRCh38
rs782317125 Chr.10: 5204382-5204382 on Build GRCh38
rs182282006 Chr.10: 5204522-5204522 on Build GRCh38
rs781832786 Chr.10: 5204508-5204509 on Build GRCh38
rs782236272 Chr.10: 5204497-5204497 on Build GRCh38
rs782525505 Chr.10: 5204459-5204459 on Build GRCh38
rs566667994 Chr.10: 5204317-5204317 on Build GRCh38
rs61730888 Chr.10: 5204399-5204399 on Build GRCh38
rs782774428 Chr.10: 5204405-5204405 on Build GRCh38
rs782244553 Chr.10: 5204336-5204336 on Build GRCh38
rs375331356 Chr.10: 5204372-5204372 on Build GRCh38
rs368974638 Chr.10: 5204473-5204473 on Build GRCh38
rs782706661 Chr.10: 5204433-5204434 on Build GRCh38

View Full Product Details