Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123761733-123762002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPXM2
Entrez Gene ID
119587
Gene Name
carboxypeptidase X (M14 family), member 2
Gene Aliases
CPX2, UNQ676
UniGene
Hs.656887
Gene Chromosome Location
Chr.10: 123745636-123944094 on Build GRCh38
Transcripts
NM_198148.2 XM_005269528.3 XM_011539283.2 XM_011539285.2 XM_011539286.1 XM_017015673.1 XM_017015674.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4786806 c.178G>T p.D60Y Chr.10: 123761959-123761959 on Build GRCh38
6518384 c.240G>T p.G80G Chr.10: 123761897-123761897 on Build GRCh38
1346539 c.1692C>T p.D564D Chr.10: 123761957-123761957 on Build GRCh38
7362567 c.1774G>T p.G592* Chr.10: 123761875-123761875 on Build GRCh38
6495606 c.263G>C p.G88A Chr.10: 123761874-123761874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776432314 Chr.10: 123761900-123761900 on Build GRCh38
rs375460068 Chr.10: 123761901-123761901 on Build GRCh38
rs943461132 Chr.10: 123761939-123761939 on Build GRCh38
rs368055115 Chr.10: 123761826-123761826 on Build GRCh38
rs754357149 Chr.10: 123761766-123761766 on Build GRCh38
rs750771582 Chr.10: 123761875-123761875 on Build GRCh38
rs780378275 Chr.10: 123761953-123761953 on Build GRCh38
rs778892334 Chr.10: 123761971-123761971 on Build GRCh38
rs909957120 Chr.10: 123761810-123761810 on Build GRCh38
rs762435610 Chr.10: 123761933-123761933 on Build GRCh38
rs372149847 Chr.10: 123761851-123761851 on Build GRCh38
rs768602178 Chr.10: 123761882-123761882 on Build GRCh38
rs779077181 Chr.10: 123761800-123761800 on Build GRCh38
rs200076195 Chr.10: 123761837-123761837 on Build GRCh38
rs535972731 Chr.10: 123761827-123761827 on Build GRCh38
rs757739744 Chr.10: 123761865-123761865 on Build GRCh38
rs529727115 Chr.10: 123761952-123761952 on Build GRCh38
rs766138240 Chr.10: 123761836-123761836 on Build GRCh38
rs374894031 Chr.10: 123761840-123761840 on Build GRCh38
rs369681113 Chr.10: 123761879-123761879 on Build GRCh38
rs187112684 Chr.10: 123761805-123761805 on Build GRCh38
rs570771178 Chr.10: 123761797-123761797 on Build GRCh38
rs773015713 Chr.10: 123761830-123761830 on Build GRCh38
rs768083943 Chr.10: 123761934-123761934 on Build GRCh38
rs549600217 Chr.10: 123761786-123761786 on Build GRCh38
rs375442985 Chr.10: 123761856-123761856 on Build GRCh38
rs760572689 Chr.10: 123761832-123761832 on Build GRCh38
rs751042921 Chr.10: 123761932-123761932 on Build GRCh38
rs533803139 Chr.10: 123761789-123761789 on Build GRCh38
rs148621668 Chr.10: 123761908-123761908 on Build GRCh38
rs564623475 Chr.10: 123761878-123761878 on Build GRCh38
rs958979117 Chr.10: 123761762-123761762 on Build GRCh38
rs746059150 Chr.10: 123761825-123761825 on Build GRCh38
rs369250317 Chr.10: 123761841-123761841 on Build GRCh38
rs750058389 Chr.10: 123761981-123761981 on Build GRCh38
rs373117160 Chr.10: 123761880-123761880 on Build GRCh38
rs372251330 Chr.10: 123761829-123761829 on Build GRCh38
rs762365746 Chr.10: 123761842-123761842 on Build GRCh38
rs547897756 Chr.10: 123761957-123761957 on Build GRCh38
rs757684763 Chr.10: 123761823-123761823 on Build GRCh38
rs748178531 Chr.10: 123761972-123761972 on Build GRCh38
rs1035865777 Chr.10: 123761858-123761858 on Build GRCh38
rs201876787 Chr.10: 123761915-123761915 on Build GRCh38
rs556355936 Chr.10: 123761807-123761807 on Build GRCh38
rs1034969724 Chr.10: 123761756-123761756 on Build GRCh38
rs753988266 Chr.10: 123761956-123761956 on Build GRCh38
rs576715653 Chr.10: 123761885-123761885 on Build GRCh38
rs552055898 Chr.10: 123761790-123761790 on Build GRCh38
rs749448782 Chr.10: 123761881-123761881 on Build GRCh38
rs746915657 Chr.10: 123761892-123761892 on Build GRCh38
rs559678940 Chr.10: 123761970-123761970 on Build GRCh38
rs763482697 Chr.10: 123761930-123761930 on Build GRCh38
rs1014928363 Chr.10: 123761784-123761784 on Build GRCh38
rs145963968 Chr.10: 123761896-123761896 on Build GRCh38
rs191938148 Chr.10: 123761819-123761819 on Build GRCh38
rs142483883 Chr.10: 123761874-123761874 on Build GRCh38
rs112981683 Chr.10: 123761919-123761919 on Build GRCh38
rs750826613 Chr.10: 123761946-123761946 on Build GRCh38

View Full Product Details