Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66469695-66469920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEAN1
Entrez Gene ID
146227
Gene Name
brain expressed, associated with NEDD4, 1
Gene Aliases
BEAN, SCA31
UniGene
Hs.740218
Gene Chromosome Location
Chr.16: 66427297-66495288 on Build GRCh38
Transcripts
NM_001136106.4 NM_001178020.2 NM_001197224.3 NM_001197225.3 XM_011522883.1 XM_011522884.1 XM_011522887.2 XM_011522888.1 XM_011522889.1 XM_011522890.1 XM_011522891.1 XM_011522892.2 XM_011522895.1 NR_109960.1

Gene Symbol
BEAN1-AS1
Entrez Gene ID
101927726
Gene Name
BEAN1 antisense RNA 1
Gene Aliases
CTD-2258A20.5
UniGene
Hs.739193
Gene Chromosome Location
Chr.16: 66469796-66476402 on Build GRCh38
Transcripts
NM_001136106.4 NM_001178020.2 NM_001197224.3 NM_001197225.3 XM_011522883.1 XM_011522884.1 XM_011522887.2 XM_011522888.1 XM_011522889.1 XM_011522890.1 XM_011522891.1 XM_011522892.2 XM_011522895.1 NR_109960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1609489 c.269G>A p.R90Q Chr.16: 66469845-66469845 on Build GRCh38
2993592 c.124G>A p.V42I Chr.16: 66469700-66469700 on Build GRCh38
6421700 c.247C>T p.H83Y Chr.16: 66469823-66469823 on Build GRCh38
6423494 c.248A>T p.H83L Chr.16: 66469824-66469824 on Build GRCh38
4650387 c.282C>T p.H94H Chr.16: 66469858-66469858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775454473 Chr.16: 66469773-66469773 on Build GRCh38
rs761750785 Chr.16: 66469844-66469844 on Build GRCh38
rs768388401 Chr.16: 66469803-66469804 on Build GRCh38
rs377695318 Chr.16: 66469774-66469774 on Build GRCh38
rs775433153 Chr.16: 66469788-66469789 on Build GRCh38
rs935234753 Chr.16: 66469717-66469717 on Build GRCh38
rs753445501 Chr.16: 66469873-66469873 on Build GRCh38
rs748550493 Chr.16: 66469772-66469772 on Build GRCh38
rs768531415 Chr.16: 66469836-66469836 on Build GRCh38
rs760169707 Chr.16: 66469858-66469858 on Build GRCh38
rs201618286 Chr.16: 66469822-66469822 on Build GRCh38
rs745370286 Chr.16: 66469783-66469784 on Build GRCh38
rs773454613 Chr.16: 66469857-66469858 on Build GRCh38
rs184299677 Chr.16: 66469781-66469781 on Build GRCh38
rs746819150 Chr.16: 66469804-66469806 on Build GRCh38
rs774258673 Chr.16: 66469839-66469839 on Build GRCh38
rs981938647 Chr.16: 66469835-66469835 on Build GRCh38
rs759861522 Chr.16: 66469809-66469809 on Build GRCh38
rs77961560 Chr.16: 66469819-66469819 on Build GRCh38
rs1042699272 Chr.16: 66469832-66469832 on Build GRCh38
rs778201206 Chr.16: 66469782-66469782 on Build GRCh38
rs773093318 Chr.16: 66469852-66469852 on Build GRCh38
rs554992323 Chr.16: 66469754-66469754 on Build GRCh38
rs973418894 Chr.16: 66469803-66469803 on Build GRCh38
rs890742299 Chr.16: 66469793-66469793 on Build GRCh38
rs200706119 Chr.16: 66469802-66469802 on Build GRCh38
rs765824418 Chr.16: 66469872-66469872 on Build GRCh38
rs986491047 Chr.16: 66469766-66469766 on Build GRCh38
rs893493147 Chr.16: 66469767-66469767 on Build GRCh38
rs747105678 Chr.16: 66469794-66469794 on Build GRCh38
rs753044229 Chr.16: 66469893-66469893 on Build GRCh38
rs564817056 Chr.16: 66469789-66469797 on Build GRCh38
rs918502280 Chr.16: 66469732-66469732 on Build GRCh38
rs72788593 Chr.16: 66469875-66469875 on Build GRCh38
rs1054189666 Chr.16: 66469788-66469788 on Build GRCh38
rs767479273 Chr.16: 66469845-66469845 on Build GRCh38
rs764916005 Chr.16: 66469876-66469876 on Build GRCh38
rs551847092 Chr.16: 66469830-66469830 on Build GRCh38
rs35852297 Chr.16: 66469744-66469744 on Build GRCh38
rs988022851 Chr.16: 66469849-66469849 on Build GRCh38
rs899575182 Chr.16: 66469786-66469786 on Build GRCh38

View Full Product Details