Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 116087428-116087667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD10
Entrez Gene ID
51397
Gene Name
COMM domain containing 10
Gene Aliases
PTD002
UniGene
Hs.483136
Gene Chromosome Location
Chr.5: 116084592-116446462 on Build GRCh38
Transcripts
NM_001308080.1 NM_016144.3 XM_011543458.2 XM_011543460.2 XM_017009553.1 XM_017009555.1 XM_017009556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4460295 c.115C>T p.Q39* Chr.5: 116087570-116087570 on Build GRCh38
4674141 c.128T>C p.L43P Chr.5: 116087583-116087583 on Build GRCh38
1432614 c.58T>C p.S20P Chr.5: 116087513-116087513 on Build GRCh38
7412061 c.53C>T p.A18V Chr.5: 116087508-116087508 on Build GRCh38
5867073 c.86G>A p.R29K Chr.5: 116087541-116087541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003525816 Chr.5: 116087620-116087620 on Build GRCh38
rs768749495 Chr.5: 116087497-116087497 on Build GRCh38
rs922840596 Chr.5: 116087551-116087551 on Build GRCh38
rs747571612 Chr.5: 116087579-116087579 on Build GRCh38
rs17138858 Chr.5: 116087457-116087457 on Build GRCh38
rs748321620 Chr.5: 116087564-116087564 on Build GRCh38
rs773722398 Chr.5: 116087511-116087511 on Build GRCh38
rs754137825 Chr.5: 116087528-116087528 on Build GRCh38
rs192150781 Chr.5: 116087612-116087612 on Build GRCh38
rs930207914 Chr.5: 116087558-116087558 on Build GRCh38
rs745630687 Chr.5: 116087560-116087560 on Build GRCh38
rs1034994411 Chr.5: 116087636-116087636 on Build GRCh38
rs766795316 Chr.5: 116087526-116087526 on Build GRCh38
rs752350926 Chr.5: 116087532-116087532 on Build GRCh38
rs149347900 Chr.5: 116087531-116087531 on Build GRCh38
rs746889941 Chr.5: 116087465-116087465 on Build GRCh38
rs74348140 Chr.5: 116087535-116087535 on Build GRCh38
rs772099312 Chr.5: 116087510-116087510 on Build GRCh38
rs144736357 Chr.5: 116087536-116087536 on Build GRCh38
rs749594188 Chr.5: 116087496-116087496 on Build GRCh38
rs775014503 Chr.5: 116087598-116087598 on Build GRCh38
rs559542713 Chr.5: 116087517-116087517 on Build GRCh38
rs944366806 Chr.5: 116087499-116087499 on Build GRCh38
rs988491361 Chr.5: 116087603-116087603 on Build GRCh38
rs370748142 Chr.5: 116087557-116087557 on Build GRCh38
rs777880617 Chr.5: 116087463-116087463 on Build GRCh38
rs367966189 Chr.5: 116087605-116087605 on Build GRCh38
rs772143963 Chr.5: 116087572-116087572 on Build GRCh38
rs548731178 Chr.5: 116087563-116087563 on Build GRCh38
rs770867874 Chr.5: 116087489-116087489 on Build GRCh38
rs181454362 Chr.5: 116087502-116087502 on Build GRCh38
rs77776961 Chr.5: 116087571-116087571 on Build GRCh38
rs761103254 Chr.5: 116087523-116087523 on Build GRCh38
rs755884450 Chr.5: 116087633-116087633 on Build GRCh38
rs369084397 Chr.5: 116087624-116087624 on Build GRCh38
rs751104947 Chr.5: 116087606-116087606 on Build GRCh38
rs774539827 Chr.5: 116087498-116087498 on Build GRCh38
rs562138361 Chr.5: 116087577-116087577 on Build GRCh38
rs768965375 Chr.5: 116087561-116087561 on Build GRCh38
rs141226616 Chr.5: 116087585-116087585 on Build GRCh38
rs747173823 Chr.5: 116087577-116087588 on Build GRCh38
rs777091905 Chr.5: 116087583-116087583 on Build GRCh38
rs201618279 Chr.5: 116087623-116087623 on Build GRCh38
rs140307563 Chr.5: 116087477-116087478 on Build GRCh38
rs771318731 Chr.5: 116087581-116087581 on Build GRCh38
rs187312352 Chr.5: 116087602-116087602 on Build GRCh38
rs888588936 Chr.5: 116087600-116087600 on Build GRCh38
rs771268578 Chr.5: 116087623-116087623 on Build GRCh38
rs765584441 Chr.5: 116087591-116087591 on Build GRCh38
rs758367744 Chr.5: 116087474-116087475 on Build GRCh38

View Full Product Details