Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151048776-151049013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf56
Entrez Gene ID
54964
Gene Name
chromosome 1 open reading frame 56
Gene Aliases
MENT
UniGene
Hs.549171
Gene Chromosome Location
Chr.1: 151047783-151051420 on Build GRCh38
Transcripts
NM_017860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1190694 c.975_976delCC p.S327fs*>15 Chr.1: 151048822-151048823 on Build GRCh38
6328672 c.987G>T p.V329V Chr.1: 151048834-151048834 on Build GRCh38
1190693 c.974_975delGC p.S325fs*>17 Chr.1: 151048821-151048822 on Build GRCh38
3667342 c.971A>G p.N324S Chr.1: 151048818-151048818 on Build GRCh38
4667412 c.1005_1005+3delAGTA p.? Chr.1: 151048852-151048855 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs969292537 Chr.1: 151048889-151048889 on Build GRCh38
rs750043639 Chr.1: 151048915-151048915 on Build GRCh38
rs761053890 Chr.1: 151048873-151048873 on Build GRCh38
rs148174196 Chr.1: 151048813-151048813 on Build GRCh38
rs587597820 Chr.1: 151048815-151048815 on Build GRCh38
rs995027811 Chr.1: 151048928-151048928 on Build GRCh38
rs934937580 Chr.1: 151048958-151048958 on Build GRCh38
rs760059277 Chr.1: 151048847-151048847 on Build GRCh38
rs757919980 Chr.1: 151048802-151048802 on Build GRCh38
rs746923017 Chr.1: 151048807-151048807 on Build GRCh38
rs375715179 Chr.1: 151048876-151048876 on Build GRCh38
rs924763002 Chr.1: 151048930-151048930 on Build GRCh38
rs187769564 Chr.1: 151048980-151048980 on Build GRCh38
rs764991918 Chr.1: 151048888-151048888 on Build GRCh38
rs978918056 Chr.1: 151048904-151048904 on Build GRCh38
rs199749484 Chr.1: 151048806-151048806 on Build GRCh38
rs773360571 Chr.1: 151048818-151048818 on Build GRCh38
rs767828529 Chr.1: 151048853-151048853 on Build GRCh38
rs756289094 Chr.1: 151048823-151048824 on Build GRCh38
rs758009996 Chr.1: 151048916-151048916 on Build GRCh38
rs57275417 Chr.1: 151048893-151048893 on Build GRCh38
rs202112151 Chr.1: 151048825-151048825 on Build GRCh38
rs748430009 Chr.1: 151048816-151048816 on Build GRCh38
rs774754518 Chr.1: 151048827-151048827 on Build GRCh38
rs1017664758 Chr.1: 151048878-151048878 on Build GRCh38
rs587672470 Chr.1: 151048955-151048955 on Build GRCh38
rs11546017 Chr.1: 151048805-151048805 on Build GRCh38

View Full Product Details