Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32169245-32169480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT1
Entrez Gene ID
10554
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases
1-AGPAT1, G15, LPAAT-alpha, LPAATA
UniGene
Hs.409230
Gene Chromosome Location
Chr.6: 32168206-32178111 on Build GRCh38
Transcripts
NM_006411.3 NM_032741.4 XM_005248805.3 XM_005248806.2 XM_011514234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
394319 c.832A>T p.K278* Chr.6: 32169296-32169296 on Build GRCh38
1443577 c.717G>A p.T239T Chr.6: 32169411-32169411 on Build GRCh38
1283311 c.766C>T p.H256Y Chr.6: 32169362-32169362 on Build GRCh38
3872938 c.694C>T p.R232W Chr.6: 32169434-32169434 on Build GRCh38
6596355 c.703C>T p.P235S Chr.6: 32169425-32169425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975811201 Chr.6: 32169433-32169433 on Build GRCh38
rs905138560 Chr.6: 32169302-32169302 on Build GRCh38
rs777896622 Chr.6: 32169284-32169284 on Build GRCh38
rs964365178 Chr.6: 32169432-32169432 on Build GRCh38
rs1002261447 Chr.6: 32169307-32169307 on Build GRCh38
rs760726399 Chr.6: 32169347-32169347 on Build GRCh38
rs775617427 Chr.6: 32169402-32169402 on Build GRCh38
rs779566214 Chr.6: 32169317-32169317 on Build GRCh38
rs751948997 Chr.6: 32169285-32169285 on Build GRCh38
rs967530858 Chr.6: 32169335-32169335 on Build GRCh38
rs763009732 Chr.6: 32169462-32169462 on Build GRCh38
rs746381833 Chr.6: 32169294-32169294 on Build GRCh38
rs765359957 Chr.6: 32169381-32169381 on Build GRCh38
rs987013114 Chr.6: 32169340-32169340 on Build GRCh38
rs769257329 Chr.6: 32169308-32169308 on Build GRCh38
rs757621954 Chr.6: 32169292-32169292 on Build GRCh38
rs781622314 Chr.6: 32169293-32169293 on Build GRCh38
rs139093368 Chr.6: 32169341-32169341 on Build GRCh38
rs773946115 Chr.6: 32169320-32169320 on Build GRCh38
rs957776755 Chr.6: 32169310-32169310 on Build GRCh38
rs1032048998 Chr.6: 32169309-32169309 on Build GRCh38
rs937993066 Chr.6: 32169411-32169411 on Build GRCh38
rs919946051 Chr.6: 32169444-32169444 on Build GRCh38
rs867669239 Chr.6: 32169389-32169389 on Build GRCh38
rs375715875 Chr.6: 32169278-32169278 on Build GRCh38
rs941591978 Chr.6: 32169275-32169275 on Build GRCh38
rs748794275 Chr.6: 32169319-32169319 on Build GRCh38
rs747965391 Chr.6: 32169328-32169328 on Build GRCh38

View Full Product Details