Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39332972-39333245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMFG
Entrez Gene ID
9535
Gene Name
glia maturation factor gamma
Gene Aliases
GMF-GAMMA
UniGene
Hs.5210
Gene Chromosome Location
Chr.19: 39328357-39336086 on Build GRCh38
Transcripts
NM_001301008.1 NM_004877.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6587638 c.200G>C p.R67T Chr.19: 39333077-39333077 on Build GRCh38
4473441 c.184C>T p.P62S Chr.19: 39333093-39333093 on Build GRCh38
4533246 c.195G>A p.Q65Q Chr.19: 39333082-39333082 on Build GRCh38
3533686 c.165G>A p.E55E Chr.19: 39333112-39333112 on Build GRCh38
5933815 c.187G>A p.E63K Chr.19: 39333090-39333090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747604476 Chr.19: 39333046-39333046 on Build GRCh38
rs768089333 Chr.19: 39333036-39333036 on Build GRCh38
rs771437043 Chr.19: 39333048-39333048 on Build GRCh38
rs888387282 Chr.19: 39333138-39333138 on Build GRCh38
rs948009242 Chr.19: 39333213-39333213 on Build GRCh38
rs1022918940 Chr.19: 39333215-39333215 on Build GRCh38
rs992458013 Chr.19: 39333174-39333174 on Build GRCh38
rs777051753 Chr.19: 39333049-39333049 on Build GRCh38
rs751712734 Chr.19: 39333079-39333079 on Build GRCh38
rs896956999 Chr.19: 39333154-39333154 on Build GRCh38
rs376843898 Chr.19: 39333090-39333090 on Build GRCh38
rs953812567 Chr.19: 39332996-39332996 on Build GRCh38
rs780598066 Chr.19: 39333046-39333047 on Build GRCh38
rs769440764 Chr.19: 39333193-39333193 on Build GRCh38
rs755909808 Chr.19: 39333096-39333096 on Build GRCh38
rs985231926 Chr.19: 39333008-39333008 on Build GRCh38
rs551580183 Chr.19: 39333056-39333056 on Build GRCh38
rs754646161 Chr.19: 39333153-39333153 on Build GRCh38
rs747706667 Chr.19: 39333088-39333088 on Build GRCh38
rs1023203370 Chr.19: 39333014-39333014 on Build GRCh38
rs1036159741 Chr.19: 39333189-39333189 on Build GRCh38
rs967784605 Chr.19: 39333039-39333039 on Build GRCh38
rs778476836 Chr.19: 39333165-39333165 on Build GRCh38
rs879780884 Chr.19: 39333220-39333220 on Build GRCh38
rs748946918 Chr.19: 39333031-39333031 on Build GRCh38
rs771561705 Chr.19: 39333170-39333170 on Build GRCh38
rs1049390911 Chr.19: 39333125-39333125 on Build GRCh38
rs369173001 Chr.19: 39333071-39333071 on Build GRCh38
rs932282367 Chr.19: 39333101-39333101 on Build GRCh38
rs765625547 Chr.19: 39333067-39333067 on Build GRCh38
rs193149106 Chr.19: 39333198-39333198 on Build GRCh38
rs771805964 Chr.19: 39333112-39333113 on Build GRCh38
rs968925437 Chr.19: 39333019-39333019 on Build GRCh38
rs747657662 Chr.19: 39333168-39333168 on Build GRCh38
rs199816793 Chr.19: 39333110-39333110 on Build GRCh38
rs777184550 Chr.19: 39333171-39333171 on Build GRCh38
rs150080208 Chr.19: 39333092-39333092 on Build GRCh38
rs767566629 Chr.19: 39333091-39333091 on Build GRCh38
rs773879884 Chr.19: 39333041-39333041 on Build GRCh38
rs941254073 Chr.19: 39333143-39333143 on Build GRCh38
rs748994750 Chr.19: 39333135-39333135 on Build GRCh38
rs779866264 Chr.19: 39333114-39333114 on Build GRCh38
rs924774986 Chr.19: 39333044-39333044 on Build GRCh38
rs764372926 Chr.19: 39333078-39333078 on Build GRCh38
rs549677229 Chr.19: 39333219-39333219 on Build GRCh38
rs773985100 Chr.19: 39333010-39333010 on Build GRCh38
rs75510098 Chr.19: 39333209-39333209 on Build GRCh38

View Full Product Details