Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58838164-58838425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYATL2
Entrez Gene ID
219970
Gene Name
glycine-N-acyltransferase like 2
Gene Aliases
BXMAS2-10, GATF-B
UniGene
Hs.254271
Gene Chromosome Location
Chr.11: 58834065-58905699 on Build GRCh38
Transcripts
NM_145016.3 XM_011544809.2 XM_017017337.1 XM_017017338.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2034879 c.163G>A p.V55I Chr.11: 58838284-58838284 on Build GRCh38
6069397 c.87C>A p.G29G Chr.11: 58838360-58838360 on Build GRCh38
6069396 c.80T>A p.V27E Chr.11: 58838367-58838367 on Build GRCh38
109846 c.162C>T p.I54I Chr.11: 58838285-58838285 on Build GRCh38
3397900 c.103A>G p.K35E Chr.11: 58838344-58838344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760735813 Chr.11: 58838380-58838380 on Build GRCh38
rs549694065 Chr.11: 58838212-58838212 on Build GRCh38
rs757932016 Chr.11: 58838332-58838332 on Build GRCh38
rs183043797 Chr.11: 58838257-58838257 on Build GRCh38
rs556865466 Chr.11: 58838333-58838333 on Build GRCh38
rs970710088 Chr.11: 58838304-58838304 on Build GRCh38
rs866153786 Chr.11: 58838242-58838242 on Build GRCh38
rs201829761 Chr.11: 58838240-58838240 on Build GRCh38
rs879189798 Chr.11: 58838393-58838393 on Build GRCh38
rs759247571 Chr.11: 58838278-58838286 on Build GRCh38
rs990854783 Chr.11: 58838288-58838288 on Build GRCh38
rs200662630 Chr.11: 58838245-58838245 on Build GRCh38
rs142894742 Chr.11: 58838372-58838372 on Build GRCh38
rs767619166 Chr.11: 58838299-58838299 on Build GRCh38
rs186979803 Chr.11: 58838274-58838274 on Build GRCh38
rs752639684 Chr.11: 58838238-58838238 on Build GRCh38
rs148535188 Chr.11: 58838359-58838359 on Build GRCh38
rs770587770 Chr.11: 58838255-58838255 on Build GRCh38
rs1049839850 Chr.11: 58838204-58838204 on Build GRCh38
rs75458416 Chr.11: 58838220-58838220 on Build GRCh38
rs775609139 Chr.11: 58838379-58838379 on Build GRCh38
rs373928451 Chr.11: 58838229-58838229 on Build GRCh38
rs771291881 Chr.11: 58838362-58838362 on Build GRCh38
rs777646908 Chr.11: 58838243-58838243 on Build GRCh38
rs376758330 Chr.11: 58838385-58838385 on Build GRCh38
rs764080145 Chr.11: 58838382-58838382 on Build GRCh38
rs779685706 Chr.11: 58838334-58838334 on Build GRCh38
rs191830464 Chr.11: 58838313-58838313 on Build GRCh38
rs755829258 Chr.11: 58838239-58838239 on Build GRCh38
rs184265394 Chr.11: 58838323-58838323 on Build GRCh38
rs578207248 Chr.11: 58838342-58838342 on Build GRCh38
rs749611824 Chr.11: 58838360-58838360 on Build GRCh38
rs763208576 Chr.11: 58838216-58838216 on Build GRCh38
rs368402125 Chr.11: 58838232-58838232 on Build GRCh38
rs201093108 Chr.11: 58838233-58838233 on Build GRCh38
rs201184601 Chr.11: 58838322-58838322 on Build GRCh38
rs773593109 Chr.11: 58838284-58838284 on Build GRCh38
rs771710384 Chr.11: 58838272-58838272 on Build GRCh38
rs763980765 Chr.11: 58838317-58838317 on Build GRCh38
rs371816018 Chr.11: 58838291-58838291 on Build GRCh38
rs368133507 Chr.11: 58838215-58838215 on Build GRCh38
rs370339254 Chr.11: 58838389-58838389 on Build GRCh38
rs373873868 Chr.11: 58838368-58838368 on Build GRCh38
rs1015287451 Chr.11: 58838234-58838234 on Build GRCh38
rs11229655 Chr.11: 58838200-58838200 on Build GRCh38
rs370375902 Chr.11: 58838219-58838219 on Build GRCh38
rs745546178 Chr.11: 58838331-58838331 on Build GRCh38
rs759549963 Chr.11: 58838293-58838293 on Build GRCh38
rs745316737 Chr.11: 58838260-58838260 on Build GRCh38
rs267603035 Chr.11: 58838275-58838275 on Build GRCh38
rs767776632 Chr.11: 58838373-58838373 on Build GRCh38
rs140753826 Chr.11: 58838285-58838285 on Build GRCh38

View Full Product Details