Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73139230-73139479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECD
Entrez Gene ID
11319
Gene Name
ecdysoneless cell cycle regulator
Gene Aliases
GCR2, HSGT1, SGT1
UniGene
Hs.631822
Gene Chromosome Location
Chr.10: 73134524-73168095 on Build GRCh38
Transcripts
NM_001135752.1 NM_001135753.1 NM_007265.2 NR_024203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1702627 c.1327G>T p.E443* Chr.10: 73139403-73139403 on Build GRCh38
4528033 c.1507G>A p.A503T Chr.10: 73139322-73139322 on Build GRCh38
2161754 c.1336C>T p.Q446* Chr.10: 73139394-73139394 on Build GRCh38
2161753 c.1435C>T p.Q479* Chr.10: 73139394-73139394 on Build GRCh38
685392 c.1401C>T p.H467H Chr.10: 73139329-73139329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779489054 Chr.10: 73139343-73139343 on Build GRCh38
rs773708177 Chr.10: 73139424-73139424 on Build GRCh38
rs762168022 Chr.10: 73139442-73139442 on Build GRCh38
rs34865820 Chr.10: 73139410-73139410 on Build GRCh38
rs756112205 Chr.10: 73139302-73139302 on Build GRCh38
rs3736517 Chr.10: 73139353-73139353 on Build GRCh38
rs958559470 Chr.10: 73139316-73139316 on Build GRCh38
rs35652947 Chr.10: 73139422-73139422 on Build GRCh38
rs759024075 Chr.10: 73139361-73139361 on Build GRCh38
rs1007524995 Chr.10: 73139428-73139428 on Build GRCh38
rs764140915 Chr.10: 73139432-73139432 on Build GRCh38
rs574287013 Chr.10: 73139400-73139400 on Build GRCh38
rs752847696 Chr.10: 73139296-73139296 on Build GRCh38
rs1036705005 Chr.10: 73139367-73139367 on Build GRCh38
rs746532921 Chr.10: 73139387-73139387 on Build GRCh38
rs763460587 Chr.10: 73139374-73139378 on Build GRCh38
rs746259542 Chr.10: 73139320-73139320 on Build GRCh38
rs758769408 Chr.10: 73139457-73139457 on Build GRCh38
rs776189860 Chr.10: 73139395-73139395 on Build GRCh38
rs372478574 Chr.10: 73139263-73139263 on Build GRCh38
rs767991255 Chr.10: 73139392-73139392 on Build GRCh38
rs772984243 Chr.10: 73139291-73139291 on Build GRCh38
rs376462298 Chr.10: 73139413-73139413 on Build GRCh38
rs747860932 Chr.10: 73139397-73139397 on Build GRCh38
rs765072176 Chr.10: 73139289-73139289 on Build GRCh38
rs765534407 Chr.10: 73139452-73139452 on Build GRCh38
rs143345132 Chr.10: 73139310-73139310 on Build GRCh38
rs373470186 Chr.10: 73139293-73139293 on Build GRCh38
rs770936832 Chr.10: 73139421-73139421 on Build GRCh38
rs3736518 Chr.10: 73139376-73139376 on Build GRCh38
rs753949042 Chr.10: 73139438-73139438 on Build GRCh38

View Full Product Details