Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75391002-75391257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR1L2
Entrez Gene ID
144321
Gene Name
GLI pathogenesis-related 1 like 2
Gene Aliases
-
UniGene
Hs.406728
Gene Chromosome Location
Chr.12: 75391070-75432688 on Build GRCh38
Transcripts
NM_001270396.1 NM_152436.2 NR_072995.1 XM_011537949.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4044843 c.136C>T p.P46S Chr.12: 75391252-75391252 on Build GRCh38
5182343 c.123C>T p.N41N Chr.12: 75391239-75391239 on Build GRCh38
5421263 c.44C>G p.S15C Chr.12: 75391160-75391160 on Build GRCh38
6287435 c.38delC p.Q14fs*7 Chr.12: 75391154-75391154 on Build GRCh38
4044838 c.56C>A p.A19E Chr.12: 75391172-75391172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915192504 Chr.12: 75391190-75391190 on Build GRCh38
rs776295229 Chr.12: 75391183-75391183 on Build GRCh38
rs572370742 Chr.12: 75391021-75391021 on Build GRCh38
rs759799456 Chr.12: 75391217-75391217 on Build GRCh38
rs927214054 Chr.12: 75391040-75391040 on Build GRCh38
rs757195299 Chr.12: 75391155-75391155 on Build GRCh38
rs962899778 Chr.12: 75391030-75391030 on Build GRCh38
rs770439759 Chr.12: 75391090-75391090 on Build GRCh38
rs892576897 Chr.12: 75391042-75391042 on Build GRCh38
rs763167969 Chr.12: 75391146-75391146 on Build GRCh38
rs927621711 Chr.12: 75391072-75391072 on Build GRCh38
rs748704522 Chr.12: 75391127-75391127 on Build GRCh38
rs142881216 Chr.12: 75391147-75391147 on Build GRCh38
rs755932910 Chr.12: 75391153-75391153 on Build GRCh38
rs533559197 Chr.12: 75391165-75391165 on Build GRCh38
rs761145902 Chr.12: 75391133-75391133 on Build GRCh38
rs144850413 Chr.12: 75391215-75391215 on Build GRCh38
rs769121895 Chr.12: 75391195-75391195 on Build GRCh38
rs200810673 Chr.12: 75391229-75391229 on Build GRCh38
rs776919229 Chr.12: 75391197-75391197 on Build GRCh38
rs1042607141 Chr.12: 75391124-75391124 on Build GRCh38
rs753788773 Chr.12: 75391154-75391154 on Build GRCh38
rs762962565 Chr.12: 75391213-75391213 on Build GRCh38
rs746701706 Chr.12: 75391179-75391179 on Build GRCh38
rs370901558 Chr.12: 75391131-75391131 on Build GRCh38
rs748808305 Chr.12: 75391088-75391088 on Build GRCh38
rs748064323 Chr.12: 75391084-75391084 on Build GRCh38
rs754772566 Chr.12: 75391081-75391081 on Build GRCh38
rs762277575 Chr.12: 75391141-75391141 on Build GRCh38
rs140903731 Chr.12: 75391210-75391210 on Build GRCh38
rs547320911 Chr.12: 75391203-75391203 on Build GRCh38
rs779886245 Chr.12: 75391178-75391178 on Build GRCh38
rs375758674 Chr.12: 75391123-75391123 on Build GRCh38
rs763860591 Chr.12: 75391221-75391221 on Build GRCh38
rs755986324 Chr.12: 75391125-75391125 on Build GRCh38
rs143963633 Chr.12: 75391064-75391064 on Build GRCh38
rs752439276 Chr.12: 75391152-75391152 on Build GRCh38
rs113853390 Chr.12: 75391045-75391045 on Build GRCh38
rs115042430 Chr.12: 75391223-75391223 on Build GRCh38
rs770440358 Chr.12: 75391128-75391128 on Build GRCh38
rs753922924 Chr.12: 75391113-75391113 on Build GRCh38
rs759148238 Chr.12: 75391099-75391099 on Build GRCh38
rs151080676 Chr.12: 75391162-75391162 on Build GRCh38
rs200745142 Chr.12: 75391085-75391085 on Build GRCh38
rs376300383 Chr.12: 75391142-75391142 on Build GRCh38
rs778332182 Chr.12: 75391130-75391130 on Build GRCh38
rs946591439 Chr.12: 75391118-75391118 on Build GRCh38
rs771090274 Chr.12: 75391161-75391161 on Build GRCh38
rs762376976 Chr.12: 75391199-75391199 on Build GRCh38
rs893707783 Chr.12: 75391116-75391116 on Build GRCh38
rs990445943 Chr.12: 75391187-75391187 on Build GRCh38
rs368593598 Chr.12: 75391126-75391126 on Build GRCh38
rs937651107 Chr.12: 75391080-75391080 on Build GRCh38
rs757391078 Chr.12: 75391114-75391114 on Build GRCh38
rs375633723 Chr.12: 75391182-75391182 on Build GRCh38
rs764714259 Chr.12: 75391111-75391111 on Build GRCh38
rs765275021 Chr.12: 75391115-75391115 on Build GRCh38
rs771804298 Chr.12: 75391168-75391168 on Build GRCh38
rs746742419 Chr.12: 75391075-75391075 on Build GRCh38
rs757366160 Chr.12: 75391066-75391066 on Build GRCh38
rs969007404 Chr.12: 75391063-75391063 on Build GRCh38
rs951165297 Chr.12: 75391101-75391101 on Build GRCh38
rs767896818 Chr.12: 75391104-75391104 on Build GRCh38
rs774080909 Chr.12: 75391093-75391093 on Build GRCh38
rs780961274 Chr.12: 75391121-75391121 on Build GRCh38
rs370742897 Chr.12: 75391166-75391166 on Build GRCh38
rs962951287 Chr.12: 75391140-75391140 on Build GRCh38
rs1009568416 Chr.12: 75391043-75391043 on Build GRCh38
rs562714429 Chr.12: 75391108-75391108 on Build GRCh38
rs937228616 Chr.12: 75391046-75391046 on Build GRCh38
rs1017221362 Chr.12: 75391139-75391139 on Build GRCh38
rs750637948 Chr.12: 75391117-75391117 on Build GRCh38
rs535323724 Chr.12: 75391134-75391134 on Build GRCh38
rs11549926 Chr.12: 75391159-75391159 on Build GRCh38
rs780899066 Chr.12: 75391083-75391083 on Build GRCh38
rs751746189 Chr.12: 75391216-75391216 on Build GRCh38
rs779220841 Chr.12: 75391070-75391070 on Build GRCh38
rs919050091 Chr.12: 75391047-75391047 on Build GRCh38

View Full Product Details