Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186652052-186652319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FETUB
Entrez Gene ID
26998
Gene Name
fetuin B
Gene Aliases
16G2, Gugu, IRL685
UniGene
Hs.81073
Gene Chromosome Location
Chr.3: 186635828-186653008 on Build GRCh38
Transcripts
NM_001308077.1 NM_001308079.1 NM_014375.2 XM_005247351.3 XM_011512681.2 XM_011512682.2 XM_011512683.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5615246 c.816C>A p.N272K Chr.3: 186652298-186652298 on Build GRCh38
5883791 c.827C>A p.T276K Chr.3: 186652309-186652309 on Build GRCh38
3365110 c.834T>G p.L278L Chr.3: 186652316-186652316 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs80348871 Chr.3: 186652086-186652086 on Build GRCh38
rs200453608 Chr.3: 186652189-186652189 on Build GRCh38
rs767803397 Chr.3: 186652133-186652133 on Build GRCh38
rs775224631 Chr.3: 186652151-186652151 on Build GRCh38
rs745492801 Chr.3: 186652250-186652250 on Build GRCh38
rs763130941 Chr.3: 186652272-186652272 on Build GRCh38
rs150847443 Chr.3: 186652147-186652147 on Build GRCh38
rs756613931 Chr.3: 186652215-186652215 on Build GRCh38
rs909867448 Chr.3: 186652192-186652192 on Build GRCh38
rs978670133 Chr.3: 186652080-186652080 on Build GRCh38
rs753015842 Chr.3: 186652213-186652213 on Build GRCh38
rs774610557 Chr.3: 186652296-186652296 on Build GRCh38
rs77190643 Chr.3: 186652158-186652158 on Build GRCh38
rs1039366369 Chr.3: 186652160-186652160 on Build GRCh38
rs766361957 Chr.3: 186652287-186652287 on Build GRCh38
rs534975364 Chr.3: 186652088-186652088 on Build GRCh38
rs756715096 Chr.3: 186652242-186652242 on Build GRCh38
rs776422401 Chr.3: 186652267-186652267 on Build GRCh38
rs748885195 Chr.3: 186652224-186652224 on Build GRCh38
rs575770663 Chr.3: 186652244-186652244 on Build GRCh38
rs778080881 Chr.3: 186652218-186652218 on Build GRCh38
rs370905116 Chr.3: 186652270-186652270 on Build GRCh38
rs976596034 Chr.3: 186652145-186652145 on Build GRCh38
rs773504499 Chr.3: 186652286-186652287 on Build GRCh38
rs771600127 Chr.3: 186652095-186652095 on Build GRCh38
rs753873366 Chr.3: 186652194-186652194 on Build GRCh38
rs768653042 Chr.3: 186652265-186652265 on Build GRCh38
rs746734763 Chr.3: 186652258-186652258 on Build GRCh38
rs181827913 Chr.3: 186652107-186652107 on Build GRCh38
rs775456964 Chr.3: 186652256-186652256 on Build GRCh38

View Full Product Details