Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75029417-75029668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUPD1
Entrez Gene ID
338599
Gene Name
dual specificity phosphatase and pro isomerase domain containing 1
Gene Aliases
DUSP27, FMDSP
UniGene
Hs.535090
Gene Chromosome Location
Chr.10: 75026427-75073827 on Build GRCh38
Transcripts
XM_011539747.2 NM_001256468.1 NM_001256469.1 NM_012330.3 XM_005269664.2 XM_017016000.1 XM_017016001.1 XM_017016002.1 XM_017016003.1 XM_017016004.1 XM_017016005.1 XM_017016006.1 XM_017016007.1 XM_017016008.1 XM_017016009.1

Gene Symbol
KAT6B
Entrez Gene ID
23522
Gene Name
lysine acetyltransferase 6B
Gene Aliases
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf
UniGene
Hs.35758
Gene Chromosome Location
Chr.10: 74824927-75032623 on Build GRCh38
Transcripts
XM_011539747.2 NM_001256468.1 NM_001256469.1 NM_012330.3 XM_005269664.2 XM_017016000.1 XM_017016001.1 XM_017016002.1 XM_017016003.1 XM_017016004.1 XM_017016005.1 XM_017016006.1 XM_017016007.1 XM_017016008.1 XM_017016009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7276815 c.4761G>T p.T1587T Chr.10: 75029585-75029585 on Build GRCh38
162774 c.4661A>G p.E1554G Chr.10: 75029485-75029485 on Build GRCh38
289833 c.4779G>A p.Q1593Q Chr.10: 75029603-75029603 on Build GRCh38
5576215 c.4622C>G p.T1541S Chr.10: 75029446-75029446 on Build GRCh38
1321197 c.4680G>T p.Q1560H Chr.10: 75029504-75029504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774479133 Chr.10: 75029614-75029614 on Build GRCh38
rs753937127 Chr.10: 75029436-75029436 on Build GRCh38
rs762428873 Chr.10: 75029603-75029603 on Build GRCh38
rs576647301 Chr.10: 75029561-75029561 on Build GRCh38
rs772986982 Chr.10: 75029493-75029493 on Build GRCh38
rs758444265 Chr.10: 75029648-75029648 on Build GRCh38
rs762953874 Chr.10: 75029496-75029496 on Build GRCh38
rs761295315 Chr.10: 75029484-75029484 on Build GRCh38
rs140322278 Chr.10: 75029562-75029562 on Build GRCh38
rs147158118 Chr.10: 75029483-75029483 on Build GRCh38
rs747832667 Chr.10: 75029589-75029589 on Build GRCh38
rs769119588 Chr.10: 75029594-75029594 on Build GRCh38
rs754132525 Chr.10: 75029553-75029553 on Build GRCh38
rs770898162 Chr.10: 75029606-75029606 on Build GRCh38
rs772255130 Chr.10: 75029456-75029456 on Build GRCh38
rs375290304 Chr.10: 75029453-75029453 on Build GRCh38
rs769370576 Chr.10: 75029492-75029492 on Build GRCh38
rs142688976 Chr.10: 75029631-75029631 on Build GRCh38
rs750507488 Chr.10: 75029644-75029644 on Build GRCh38
rs765433545 Chr.10: 75029640-75029640 on Build GRCh38
rs145636342 Chr.10: 75029584-75029584 on Build GRCh38
rs762105075 Chr.10: 75029639-75029639 on Build GRCh38
rs749038773 Chr.10: 75029490-75029492 on Build GRCh38
rs751387257 Chr.10: 75029526-75029526 on Build GRCh38
rs377200752 Chr.10: 75029630-75029630 on Build GRCh38
rs372545959 Chr.10: 75029585-75029585 on Build GRCh38
rs149654793 Chr.10: 75029582-75029582 on Build GRCh38
rs747427953 Chr.10: 75029471-75029471 on Build GRCh38
rs766030818 Chr.10: 75029513-75029513 on Build GRCh38
rs779236437 Chr.10: 75029447-75029447 on Build GRCh38
rs959080642 Chr.10: 75029521-75029521 on Build GRCh38
rs145158232 Chr.10: 75029457-75029457 on Build GRCh38
rs768290031 Chr.10: 75029480-75029480 on Build GRCh38
rs757461393 Chr.10: 75029441-75029441 on Build GRCh38
rs148492740 Chr.10: 75029600-75029600 on Build GRCh38
rs368252918 Chr.10: 75029551-75029551 on Build GRCh38
rs757242396 Chr.10: 75029554-75029554 on Build GRCh38
rs758921740 Chr.10: 75029574-75029574 on Build GRCh38

View Full Product Details