Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49251671-49251896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP3
Entrez Gene ID
50943
Gene Name
forkhead box P3
Gene Aliases
AIID, DIETER, IPEX, JM2, PIDX, XPID
UniGene
Hs.247700
Gene Chromosome Location
Chr.X: 49250436-49266505 on Build GRCh38
Transcripts
NM_001114377.1 NM_014009.3 XM_006724533.2 XM_011543916.2 XM_017029565.1 XM_017029566.1 XM_017029567.1

SNP Detail

SNP ID Chromosome Location
rs782695804 Chr.X: 49251856-49251856 on Build GRCh38
rs781915091 Chr.X: 49251792-49251792 on Build GRCh38
rs1022817617 Chr.X: 49251844-49251844 on Build GRCh38
rs1057518281 Chr.X: 49251702-49251702 on Build GRCh38
rs782127412 Chr.X: 49251759-49251759 on Build GRCh38
rs782012008 Chr.X: 49251774-49251775 on Build GRCh38
rs968640529 Chr.X: 49251857-49251857 on Build GRCh38
rs374337927 Chr.X: 49251814-49251814 on Build GRCh38
rs886821043 Chr.X: 49251709-49251709 on Build GRCh38
rs782162095 Chr.X: 49251825-49251825 on Build GRCh38
rs782208834 Chr.X: 49251779-49251779 on Build GRCh38
rs782320012 Chr.X: 49251785-49251785 on Build GRCh38
rs781966644 Chr.X: 49251778-49251778 on Build GRCh38
rs782289093 Chr.X: 49251811-49251812 on Build GRCh38
rs959911946 Chr.X: 49251836-49251836 on Build GRCh38
rs782321278 Chr.X: 49251768-49251768 on Build GRCh38
rs991759984 Chr.X: 49251841-49251841 on Build GRCh38
rs74162068 Chr.X: 49251776-49251776 on Build GRCh38
rs371124215 Chr.X: 49251795-49251795 on Build GRCh38
rs122467169 Chr.X: 49251698-49251698 on Build GRCh38
rs122467172 Chr.X: 49251692-49251693 on Build GRCh38
rs782443295 Chr.X: 49251704-49251704 on Build GRCh38
rs122467175 Chr.X: 49251711-49251711 on Build GRCh38
rs782790058 Chr.X: 49251799-49251799 on Build GRCh38
rs782684613 Chr.X: 49251748-49251748 on Build GRCh38

View Full Product Details