Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 50986515-50986780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAC1
Entrez Gene ID
55520
Gene Name
elaC ribonuclease Z 1
Gene Aliases
D29
UniGene
Hs.657360
Gene Chromosome Location
Chr.18: 50968017-50988120 on Build GRCh38
Transcripts
NM_018696.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1750544 c.631C>G p.P211A Chr.18: 50986624-50986624 on Build GRCh38
6297119 c.712G>T p.V238F Chr.18: 50986705-50986705 on Build GRCh38
4072501 c.703C>T p.P235S Chr.18: 50986696-50986696 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1025603587 Chr.18: 50986581-50986581 on Build GRCh38
rs769332518 Chr.18: 50986761-50986761 on Build GRCh38
rs907143744 Chr.18: 50986621-50986621 on Build GRCh38
rs998181005 Chr.18: 50986577-50986577 on Build GRCh38
rs764289320 Chr.18: 50986565-50986565 on Build GRCh38
rs200131575 Chr.18: 50986690-50986690 on Build GRCh38
rs772778327 Chr.18: 50986608-50986608 on Build GRCh38
rs991051960 Chr.18: 50986740-50986740 on Build GRCh38
rs951349362 Chr.18: 50986629-50986629 on Build GRCh38
rs751444029 Chr.18: 50986630-50986630 on Build GRCh38
rs749831260 Chr.18: 50986722-50986722 on Build GRCh38
rs184044684 Chr.18: 50986625-50986625 on Build GRCh38
rs767119821 Chr.18: 50986587-50986587 on Build GRCh38
rs753725409 Chr.18: 50986666-50986666 on Build GRCh38
rs757465081 Chr.18: 50986688-50986688 on Build GRCh38
rs569400237 Chr.18: 50986599-50986599 on Build GRCh38
rs765039487 Chr.18: 50986683-50986683 on Build GRCh38
rs201796024 Chr.18: 50986710-50986710 on Build GRCh38
rs752537627 Chr.18: 50986684-50986684 on Build GRCh38
rs754730073 Chr.18: 50986670-50986670 on Build GRCh38
rs774094372 Chr.18: 50986570-50986570 on Build GRCh38
rs1016117036 Chr.18: 50986641-50986641 on Build GRCh38
rs761531934 Chr.18: 50986573-50986573 on Build GRCh38
rs755351278 Chr.18: 50986665-50986667 on Build GRCh38
rs745901121 Chr.18: 50986701-50986701 on Build GRCh38
rs760162839 Chr.18: 50986640-50986640 on Build GRCh38
rs766285345 Chr.18: 50986660-50986660 on Build GRCh38
rs945718401 Chr.18: 50986572-50986572 on Build GRCh38

View Full Product Details