Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 100140603-100140836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO43
Entrez Gene ID
286151
Gene Name
F-box protein 43
Gene Aliases
EMI2, ERP1, FBX43
UniGene
Hs.339577
Gene Chromosome Location
Chr.8: 100133360-100150569 on Build GRCh38
Transcripts
NM_001029860.3 NR_036491.1 XM_011516987.2 XM_011516989.2 XM_011516990.1 XM_011516992.2 XM_017013358.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5522677 c.1547C>T p.S516F Chr.8: 100140707-100140707 on Build GRCh38
4564920 c.1547_1548CC>TT p.S516F Chr.8: 100140706-100140707 on Build GRCh38
5522680 c.1548C>T p.S516S Chr.8: 100140706-100140706 on Build GRCh38
7142042 c.1528C>A p.L510I Chr.8: 100140726-100140726 on Build GRCh38
4162555 c.1442A>T p.E481V Chr.8: 100140710-100140710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947812445 Chr.8: 100140691-100140691 on Build GRCh38
rs780135201 Chr.8: 100140779-100140779 on Build GRCh38
rs762500119 Chr.8: 100140698-100140698 on Build GRCh38
rs779324623 Chr.8: 100140750-100140750 on Build GRCh38
rs199780921 Chr.8: 100140700-100140700 on Build GRCh38
rs758585591 Chr.8: 100140763-100140763 on Build GRCh38
rs747863236 Chr.8: 100140780-100140780 on Build GRCh38
rs1460934 Chr.8: 100140796-100140796 on Build GRCh38
rs754292677 Chr.8: 100140731-100140731 on Build GRCh38
rs757602569 Chr.8: 100140638-100140638 on Build GRCh38
rs772840079 Chr.8: 100140688-100140688 on Build GRCh38
rs375949319 Chr.8: 100140699-100140699 on Build GRCh38
rs267601676 Chr.8: 100140761-100140761 on Build GRCh38
rs768363063 Chr.8: 100140639-100140639 on Build GRCh38
rs894232097 Chr.8: 100140756-100140756 on Build GRCh38
rs140293795 Chr.8: 100140658-100140658 on Build GRCh38
rs757678766 Chr.8: 100140745-100140745 on Build GRCh38
rs764369802 Chr.8: 100140722-100140722 on Build GRCh38
rs764936497 Chr.8: 100140687-100140687 on Build GRCh38
rs201016433 Chr.8: 100140685-100140685 on Build GRCh38
rs760623593 Chr.8: 100140637-100140637 on Build GRCh38
rs746938307 Chr.8: 100140783-100140783 on Build GRCh38
rs1027188595 Chr.8: 100140806-100140806 on Build GRCh38
rs372309665 Chr.8: 100140751-100140751 on Build GRCh38

View Full Product Details