Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76270327-76270570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBALD2
Entrez Gene ID
283991
Gene Name
UBA like domain containing 2
Gene Aliases
FAM100B
UniGene
Hs.744981
Gene Chromosome Location
Chr.17: 76265205-76271298 on Build GRCh38
Transcripts
NM_182565.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391103 c.355A>C p.S119R Chr.17: 76270365-76270365 on Build GRCh38
5959230 c.365C>T p.A122V Chr.17: 76270375-76270375 on Build GRCh38
6189976 c.344C>T p.S115L Chr.17: 76270354-76270354 on Build GRCh38
4629130 c.476C>T p.A159V Chr.17: 76270486-76270486 on Build GRCh38
6839547 c.418C>T p.Q140* Chr.17: 76270428-76270428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050990489 Chr.17: 76270463-76270463 on Build GRCh38
rs12945355 Chr.17: 76270379-76270379 on Build GRCh38
rs367907656 Chr.17: 76270361-76270361 on Build GRCh38
rs756767078 Chr.17: 76270375-76270375 on Build GRCh38
rs764676310 Chr.17: 76270376-76270376 on Build GRCh38
rs970984635 Chr.17: 76270386-76270386 on Build GRCh38
rs559457261 Chr.17: 76270513-76270513 on Build GRCh38
rs924010202 Chr.17: 76270493-76270493 on Build GRCh38
rs113424826 Chr.17: 76270516-76270516 on Build GRCh38
rs868544425 Chr.17: 76270360-76270360 on Build GRCh38
rs746384823 Chr.17: 76270484-76270484 on Build GRCh38
rs758262682 Chr.17: 76270378-76270378 on Build GRCh38
rs778030106 Chr.17: 76270392-76270392 on Build GRCh38
rs942923552 Chr.17: 76270520-76270520 on Build GRCh38
rs372144235 Chr.17: 76270354-76270354 on Build GRCh38
rs375591681 Chr.17: 76270388-76270388 on Build GRCh38
rs774534489 Chr.17: 76270435-76270435 on Build GRCh38
rs545682018 Chr.17: 76270494-76270494 on Build GRCh38
rs1044882 Chr.17: 76270514-76270514 on Build GRCh38
rs756361862 Chr.17: 76270391-76270391 on Build GRCh38
rs772650700 Chr.17: 76270488-76270488 on Build GRCh38
rs771068035 Chr.17: 76270400-76270400 on Build GRCh38
rs940674843 Chr.17: 76270399-76270399 on Build GRCh38
rs753329299 Chr.17: 76270364-76270364 on Build GRCh38
rs923321889 Chr.17: 76270420-76270420 on Build GRCh38
rs577063430 Chr.17: 76270398-76270398 on Build GRCh38
rs746802695 Chr.17: 76270390-76270390 on Build GRCh38
rs750388667 Chr.17: 76270377-76270377 on Build GRCh38
rs749379902 Chr.17: 76270395-76270395 on Build GRCh38
rs867929939 Chr.17: 76270359-76270359 on Build GRCh38

View Full Product Details