Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127945692-127945927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM102A
Entrez Gene ID
399665
Gene Name
family with sequence similarity 102 member A
Gene Aliases
C9orf132, EEIG1, SYM-3A, bA203J24.7
UniGene
Hs.535972
Gene Chromosome Location
Chr.9: 127940582-127980533 on Build GRCh38
Transcripts
NM_001035254.2 NM_203305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6392466 c.647C>T p.S216F Chr.9: 127945723-127945723 on Build GRCh38
6584831 c.671C>G p.S224C Chr.9: 127945699-127945699 on Build GRCh38
6244341 c.648C>A p.S216S Chr.9: 127945722-127945722 on Build GRCh38
4650101 c.639G>A p.Q213Q Chr.9: 127945731-127945731 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs996061573 Chr.9: 127945825-127945825 on Build GRCh38
rs777168975 Chr.9: 127945791-127945791 on Build GRCh38
rs766909257 Chr.9: 127945718-127945718 on Build GRCh38
rs749012493 Chr.9: 127945749-127945749 on Build GRCh38
rs759931225 Chr.9: 127945792-127945792 on Build GRCh38
rs76200605 Chr.9: 127945827-127945827 on Build GRCh38
rs573905860 Chr.9: 127945753-127945753 on Build GRCh38
rs565333176 Chr.9: 127945824-127945824 on Build GRCh38
rs750158933 Chr.9: 127945719-127945719 on Build GRCh38
rs1057496112 Chr.9: 127945861-127945861 on Build GRCh38
rs754935561 Chr.9: 127945751-127945751 on Build GRCh38
rs890028611 Chr.9: 127945846-127945846 on Build GRCh38
rs771219611 Chr.9: 127945783-127945783 on Build GRCh38
rs773158983 Chr.9: 127945778-127945778 on Build GRCh38
rs367780713 Chr.9: 127945731-127945731 on Build GRCh38
rs747314019 Chr.9: 127945779-127945779 on Build GRCh38
rs895822710 Chr.9: 127945868-127945868 on Build GRCh38
rs1018607403 Chr.9: 127945737-127945737 on Build GRCh38
rs200794445 Chr.9: 127945764-127945764 on Build GRCh38
rs531676668 Chr.9: 127945810-127945810 on Build GRCh38
rs138186676 Chr.9: 127945819-127945819 on Build GRCh38
rs377240122 Chr.9: 127945889-127945889 on Build GRCh38
rs770263026 Chr.9: 127945802-127945802 on Build GRCh38
rs561247510 Chr.9: 127945736-127945736 on Build GRCh38
rs200475137 Chr.9: 127945765-127945765 on Build GRCh38
rs931496641 Chr.9: 127945811-127945811 on Build GRCh38

View Full Product Details