Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28882098-28882339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLOT2
Entrez Gene ID
2319
Gene Name
flotillin 2
Gene Aliases
ECS-1, ECS1, ESA, ESA1, M17S1
UniGene
Hs.514038
Gene Chromosome Location
Chr.17: 28879339-28897728 on Build GRCh38
Transcripts
NM_004475.2 XM_005257950.3 XM_005257953.2 XM_017024394.1 XM_017024395.1 XM_017024396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3958144 c.699+1G>T p.? Chr.17: 28882117-28882117 on Build GRCh38
6080755 c.597G>A p.K199K Chr.17: 28882220-28882220 on Build GRCh38
4751616 c.615G>T p.K205N Chr.17: 28882202-28882202 on Build GRCh38
6665148 c.674C>T p.A225V Chr.17: 28882143-28882143 on Build GRCh38
2801105 c.577C>T p.R193W Chr.17: 28882339-28882339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369574598 Chr.17: 28882296-28882296 on Build GRCh38
rs751775843 Chr.17: 28882167-28882167 on Build GRCh38
rs61730542 Chr.17: 28882229-28882229 on Build GRCh38
rs370375368 Chr.17: 28882195-28882195 on Build GRCh38
rs551762020 Chr.17: 28882196-28882196 on Build GRCh38
rs1030564351 Chr.17: 28882226-28882226 on Build GRCh38
rs768948854 Chr.17: 28882210-28882210 on Build GRCh38
rs534201774 Chr.17: 28882151-28882151 on Build GRCh38
rs373765414 Chr.17: 28882282-28882282 on Build GRCh38
rs776183781 Chr.17: 28882253-28882253 on Build GRCh38
rs763916891 Chr.17: 28882283-28882283 on Build GRCh38
rs377184819 Chr.17: 28882169-28882169 on Build GRCh38
rs754118336 Chr.17: 28882137-28882137 on Build GRCh38
rs201809072 Chr.17: 28882291-28882291 on Build GRCh38
rs892488771 Chr.17: 28882157-28882157 on Build GRCh38
rs1001284609 Chr.17: 28882264-28882265 on Build GRCh38
rs752528422 Chr.17: 28882274-28882274 on Build GRCh38
rs760314503 Chr.17: 28882230-28882230 on Build GRCh38
rs750488943 Chr.17: 28882307-28882307 on Build GRCh38
rs751470834 Chr.17: 28882290-28882290 on Build GRCh38
rs764835661 Chr.17: 28882269-28882269 on Build GRCh38
rs757325912 Chr.17: 28882159-28882159 on Build GRCh38
rs770625552 Chr.17: 28882239-28882239 on Build GRCh38
rs953788381 Chr.17: 28882227-28882227 on Build GRCh38
rs745997983 Chr.17: 28882194-28882194 on Build GRCh38
rs34442762 Chr.17: 28882305-28882305 on Build GRCh38
rs756094041 Chr.17: 28882317-28882317 on Build GRCh38
rs781419681 Chr.17: 28882192-28882192 on Build GRCh38
rs777735240 Chr.17: 28882165-28882165 on Build GRCh38
rs986413774 Chr.17: 28882263-28882263 on Build GRCh38
rs749820039 Chr.17: 28882205-28882205 on Build GRCh38
rs758088150 Chr.17: 28882161-28882161 on Build GRCh38
rs759007329 Chr.17: 28882261-28882261 on Build GRCh38
rs774700368 Chr.17: 28882214-28882214 on Build GRCh38

View Full Product Details