Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20747474-20747709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDDM3A
Entrez Gene ID
10876
Gene Name
epididymal protein 3A
Gene Aliases
EP3A, FAM12A, HE3-ALPHA, HE3A, HE3ALPHA, RAM1
UniGene
Hs.304757
Gene Chromosome Location
Chr.14: 20735946-20748380 on Build GRCh38
Transcripts
NM_006683.4 XM_017020934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4877706 c.129A>G p.R43R Chr.14: 20747709-20747709 on Build GRCh38
7428471 c.30A>T p.I10I Chr.14: 20747610-20747610 on Build GRCh38
5382031 c.99C>T p.F33F Chr.14: 20747679-20747679 on Build GRCh38
954296 c.107T>G p.L36R Chr.14: 20747687-20747687 on Build GRCh38
3955883 c.110A>G p.H37R Chr.14: 20747690-20747690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140958213 Chr.14: 20747587-20747587 on Build GRCh38
rs116985083 Chr.14: 20747674-20747674 on Build GRCh38
rs561148669 Chr.14: 20747558-20747558 on Build GRCh38
rs755685949 Chr.14: 20747607-20747607 on Build GRCh38
rs754417674 Chr.14: 20747588-20747588 on Build GRCh38
rs759760624 Chr.14: 20747572-20747572 on Build GRCh38
rs778783465 Chr.14: 20747663-20747663 on Build GRCh38
rs769871623 Chr.14: 20747563-20747563 on Build GRCh38
rs772227676 Chr.14: 20747581-20747581 on Build GRCh38
rs777345529 Chr.14: 20747656-20747656 on Build GRCh38
rs777138112 Chr.14: 20747611-20747611 on Build GRCh38
rs371612511 Chr.14: 20747568-20747568 on Build GRCh38
rs768838642 Chr.14: 20747636-20747636 on Build GRCh38
rs755721035 Chr.14: 20747651-20747651 on Build GRCh38
rs371729522 Chr.14: 20747595-20747595 on Build GRCh38
rs752443739 Chr.14: 20747650-20747650 on Build GRCh38
rs10136680 Chr.14: 20747533-20747533 on Build GRCh38
rs773157447 Chr.14: 20747565-20747565 on Build GRCh38
rs1022175379 Chr.14: 20747654-20747654 on Build GRCh38
rs369884461 Chr.14: 20747599-20747599 on Build GRCh38
rs374505218 Chr.14: 20747603-20747603 on Build GRCh38
rs201332362 Chr.14: 20747589-20747589 on Build GRCh38
rs775569993 Chr.14: 20747586-20747586 on Build GRCh38
rs10135976 Chr.14: 20747569-20747569 on Build GRCh38
rs765699364 Chr.14: 20747592-20747592 on Build GRCh38
rs139377155 Chr.14: 20747646-20747646 on Build GRCh38
rs779125121 Chr.14: 20747543-20747543 on Build GRCh38
rs746716185 Chr.14: 20747555-20747555 on Build GRCh38
rs146899571 Chr.14: 20747643-20747643 on Build GRCh38
rs752282565 Chr.14: 20747606-20747606 on Build GRCh38
rs753231505 Chr.14: 20747657-20747657 on Build GRCh38
rs113735065 Chr.14: 20747533-20747533 on Build GRCh38
rs999165086 Chr.14: 20747667-20747667 on Build GRCh38
rs778786328 Chr.14: 20747618-20747618 on Build GRCh38
rs75387363 Chr.14: 20747559-20747559 on Build GRCh38
rs745615813 Chr.14: 20747625-20747625 on Build GRCh38
rs986969345 Chr.14: 20747526-20747526 on Build GRCh38
rs775729381 Chr.14: 20747631-20747631 on Build GRCh38
rs754690133 Chr.14: 20747598-20747598 on Build GRCh38
rs200261229 Chr.14: 20747662-20747662 on Build GRCh38
rs201725458 Chr.14: 20747649-20747649 on Build GRCh38
rs900323684 Chr.14: 20747639-20747639 on Build GRCh38
rs760866119 Chr.14: 20747632-20747632 on Build GRCh38
rs758157589 Chr.14: 20747613-20747613 on Build GRCh38
rs770709746 Chr.14: 20747616-20747616 on Build GRCh38
rs772313833 Chr.14: 20747628-20747628 on Build GRCh38
rs776569220 Chr.14: 20747637-20747637 on Build GRCh38

View Full Product Details