Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45890448-45890689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSA
Entrez Gene ID
5476
Gene Name
cathepsin A
Gene Aliases
GLB2, GSL, NGBE, PPCA, PPGB
UniGene
Hs.609336
Gene Chromosome Location
Chr.20: 45890144-45898820 on Build GRCh38
Transcripts
NM_000308.3 NM_001127695.2 NM_001167594.2 NR_133656.1 NM_001278535.1 NM_080749.3

Gene Symbol
NEURL2
Entrez Gene ID
140825
Gene Name
neuralized E3 ubiquitin protein ligase 2
Gene Aliases
C20orf163, OZZ, OZZ-E3
UniGene
Hs.517094
Gene Chromosome Location
Chr.20: 45888472-45891287 on Build GRCh38
Transcripts
NM_000308.3 NM_001127695.2 NM_001167594.2 NR_133656.1 NM_001278535.1 NM_080749.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1615666 c.443G>A p.R148H Chr.20: 45890549-45890549 on Build GRCh38
4603412 c.407_408insA p.S136fs*15 Chr.20: 45890584-45890585 on Build GRCh38
3293964 c.306C>A p.P102P Chr.20: 45890686-45890686 on Build GRCh38
1412101 c.491G>A p.R164Q Chr.20: 45890501-45890501 on Build GRCh38
6240910 c.395C>A p.A132E Chr.20: 45890597-45890597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758403417 Chr.20: 45890518-45890518 on Build GRCh38
rs761575367 Chr.20: 45890590-45890590 on Build GRCh38
rs781712783 Chr.20: 45890544-45890544 on Build GRCh38
rs765561816 Chr.20: 45890593-45890593 on Build GRCh38
rs925536240 Chr.20: 45890567-45890567 on Build GRCh38
rs751615450 Chr.20: 45890610-45890610 on Build GRCh38
rs1030456894 Chr.20: 45890540-45890540 on Build GRCh38
rs768542235 Chr.20: 45890574-45890574 on Build GRCh38
rs748488415 Chr.20: 45890545-45890545 on Build GRCh38
rs997216847 Chr.20: 45890520-45890520 on Build GRCh38
rs201802475 Chr.20: 45890585-45890586 on Build GRCh38
rs949488083 Chr.20: 45890623-45890623 on Build GRCh38
rs769677090 Chr.20: 45890539-45890539 on Build GRCh38
rs750566384 Chr.20: 45890515-45890515 on Build GRCh38
rs6032577 Chr.20: 45890635-45890635 on Build GRCh38
rs201162863 Chr.20: 45890547-45890547 on Build GRCh38
rs998666789 Chr.20: 45890495-45890495 on Build GRCh38
rs374500006 Chr.20: 45890566-45890566 on Build GRCh38
rs961855068 Chr.20: 45890490-45890490 on Build GRCh38
rs370061686 Chr.20: 45890601-45890601 on Build GRCh38
rs757016726 Chr.20: 45890550-45890550 on Build GRCh38
rs755490733 Chr.20: 45890535-45890535 on Build GRCh38
rs771490340 Chr.20: 45890565-45890565 on Build GRCh38
rs763893268 Chr.20: 45890475-45890475 on Build GRCh38
rs773310703 Chr.20: 45890594-45890594 on Build GRCh38
rs753583076 Chr.20: 45890492-45890492 on Build GRCh38
rs768110929 Chr.20: 45890625-45890625 on Build GRCh38
rs762055789 Chr.20: 45890508-45890508 on Build GRCh38
rs752019187 Chr.20: 45890533-45890533 on Build GRCh38
rs913887091 Chr.20: 45890611-45890611 on Build GRCh38
rs368146968 Chr.20: 45890667-45890667 on Build GRCh38
rs776450846 Chr.20: 45890576-45890576 on Build GRCh38
rs745453673 Chr.20: 45890562-45890562 on Build GRCh38
rs755579511 Chr.20: 45890622-45890622 on Build GRCh38
rs766414314 Chr.20: 45890532-45890532 on Build GRCh38
rs778420296 Chr.20: 45890557-45890557 on Build GRCh38
rs745522640 Chr.20: 45890658-45890658 on Build GRCh38
rs1031476739 Chr.20: 45890500-45890500 on Build GRCh38
rs764746127 Chr.20: 45890635-45890644 on Build GRCh38
rs765415639 Chr.20: 45890509-45890509 on Build GRCh38
rs756546707 Chr.20: 45890633-45890633 on Build GRCh38
rs753167327 Chr.20: 45890629-45890629 on Build GRCh38

View Full Product Details