Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41909644-41909915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD4
Entrez Gene ID
30844
Gene Name
EH domain containing 4
Gene Aliases
PAST4
UniGene
Hs.143703
Gene Chromosome Location
Chr.15: 41899440-41972557 on Build GRCh38
Transcripts
NM_139265.3 XM_017022100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1372878 c.1034T>C p.L345P Chr.15: 41909754-41909754 on Build GRCh38
6652448 c.1059A>G p.A353A Chr.15: 41909729-41909729 on Build GRCh38
6303626 c.925-6C>T p.? Chr.15: 41909869-41909869 on Build GRCh38
5422720 c.1010G>A p.R337K Chr.15: 41909778-41909778 on Build GRCh38
6215998 c.1049A>G p.Q350R Chr.15: 41909739-41909739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746158680 Chr.15: 41909851-41909851 on Build GRCh38
rs369508545 Chr.15: 41909771-41909771 on Build GRCh38
rs1011389905 Chr.15: 41909806-41909806 on Build GRCh38
rs757115202 Chr.15: 41909738-41909738 on Build GRCh38
rs76085783 Chr.15: 41909674-41909674 on Build GRCh38
rs766436418 Chr.15: 41909775-41909775 on Build GRCh38
rs754537007 Chr.15: 41909790-41909790 on Build GRCh38
rs778098045 Chr.15: 41909794-41909796 on Build GRCh38
rs771459487 Chr.15: 41909731-41909731 on Build GRCh38
rs758222613 Chr.15: 41909774-41909774 on Build GRCh38
rs773502722 Chr.15: 41909728-41909728 on Build GRCh38
rs776861009 Chr.15: 41909879-41909879 on Build GRCh38
rs777446463 Chr.15: 41909846-41909846 on Build GRCh38
rs890501 Chr.15: 41909765-41909765 on Build GRCh38
rs1045767795 Chr.15: 41909829-41909829 on Build GRCh38
rs780627336 Chr.15: 41909798-41909798 on Build GRCh38
rs759599419 Chr.15: 41909887-41909887 on Build GRCh38
rs370059543 Chr.15: 41909685-41909685 on Build GRCh38
rs772618529 Chr.15: 41909875-41909875 on Build GRCh38
rs767520779 Chr.15: 41909743-41909743 on Build GRCh38
rs201859741 Chr.15: 41909748-41909748 on Build GRCh38
rs181241129 Chr.15: 41909673-41909673 on Build GRCh38
rs145224561 Chr.15: 41909801-41909801 on Build GRCh38
rs201469022 Chr.15: 41909865-41909865 on Build GRCh38
rs759627626 Chr.15: 41909740-41909740 on Build GRCh38
rs755766198 Chr.15: 41909833-41909833 on Build GRCh38
rs139710378 Chr.15: 41909720-41909720 on Build GRCh38
rs778551029 Chr.15: 41909825-41909825 on Build GRCh38
rs765007322 Chr.15: 41909682-41909682 on Build GRCh38
rs1045389801 Chr.15: 41909701-41909701 on Build GRCh38
rs767749734 Chr.15: 41909670-41909670 on Build GRCh38
rs747705937 Chr.15: 41909883-41909883 on Build GRCh38
rs754484073 Chr.15: 41909714-41909714 on Build GRCh38
rs747686056 Chr.15: 41909718-41909718 on Build GRCh38
rs746443044 Chr.15: 41909870-41909870 on Build GRCh38
rs376398672 Chr.15: 41909663-41909663 on Build GRCh38
rs749677247 Chr.15: 41909849-41909849 on Build GRCh38
rs780107549 Chr.15: 41909692-41909692 on Build GRCh38
rs201399185 Chr.15: 41909880-41909880 on Build GRCh38
rs749637424 Chr.15: 41909730-41909730 on Build GRCh38
rs879454396 Chr.15: 41909737-41909737 on Build GRCh38
rs751481417 Chr.15: 41909783-41909783 on Build GRCh38
rs774845735 Chr.15: 41909866-41909866 on Build GRCh38
rs1001465092 Chr.15: 41909749-41909749 on Build GRCh38
rs746544723 Chr.15: 41909697-41909697 on Build GRCh38
rs760862597 Chr.15: 41909758-41909758 on Build GRCh38
rs758458153 Chr.15: 41909689-41909689 on Build GRCh38
rs780907907 Chr.15: 41909715-41909715 on Build GRCh38
rs750224589 Chr.15: 41909772-41909772 on Build GRCh38
rs760777718 Chr.15: 41909878-41909878 on Build GRCh38

View Full Product Details