Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72195356-72195617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLR1
Entrez Gene ID
2348
Gene Name
folate receptor 1
Gene Aliases
FBP, FOLR
UniGene
Hs.73769
Gene Chromosome Location
Chr.11: 72189558-72196323 on Build GRCh38
Transcripts
NM_000802.3 NM_016724.2 NM_016725.2 NM_016729.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5816935 c.310A>T p.T104S Chr.11: 72195412-72195412 on Build GRCh38
5839541 c.347G>A p.W116* Chr.11: 72195449-72195449 on Build GRCh38
3452978 c.268G>A p.G90R Chr.11: 72195370-72195370 on Build GRCh38
5003234 c.278C>A p.A93E Chr.11: 72195380-72195380 on Build GRCh38
5631711 c.279A>C p.A93A Chr.11: 72195381-72195381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765873354 Chr.11: 72195585-72195585 on Build GRCh38
rs764420714 Chr.11: 72195395-72195395 on Build GRCh38
rs763433361 Chr.11: 72195494-72195494 on Build GRCh38
rs753777743 Chr.11: 72195587-72195587 on Build GRCh38
rs751220498 Chr.11: 72195410-72195410 on Build GRCh38
rs759712157 Chr.11: 72195383-72195383 on Build GRCh38
rs896520206 Chr.11: 72195548-72195548 on Build GRCh38
rs766989831 Chr.11: 72195413-72195413 on Build GRCh38
rs267603174 Chr.11: 72195406-72195406 on Build GRCh38
rs749242260 Chr.11: 72195445-72195445 on Build GRCh38
rs547880612 Chr.11: 72195492-72195492 on Build GRCh38
rs756942461 Chr.11: 72195412-72195412 on Build GRCh38
rs774626222 Chr.11: 72195561-72195561 on Build GRCh38
rs765796999 Chr.11: 72195384-72195384 on Build GRCh38
rs76191655 Chr.11: 72195394-72195394 on Build GRCh38
rs121918405 Chr.11: 72195454-72195454 on Build GRCh38
rs1006659128 Chr.11: 72195380-72195380 on Build GRCh38
rs376293431 Chr.11: 72195493-72195493 on Build GRCh38
rs760166995 Chr.11: 72195579-72195579 on Build GRCh38
rs149273197 Chr.11: 72195480-72195480 on Build GRCh38
rs997891200 Chr.11: 72195431-72195431 on Build GRCh38
rs201325534 Chr.11: 72195479-72195479 on Build GRCh38
rs958812683 Chr.11: 72195489-72195489 on Build GRCh38
rs749885216 Chr.11: 72195414-72195414 on Build GRCh38
rs767114329 Chr.11: 72195569-72195569 on Build GRCh38
rs112834048 Chr.11: 72195487-72195487 on Build GRCh38
rs1047972742 Chr.11: 72195584-72195584 on Build GRCh38
rs368803339 Chr.11: 72195483-72195483 on Build GRCh38
rs1057518027 Chr.11: 72195460-72195460 on Build GRCh38
rs754798966 Chr.11: 72195477-72195477 on Build GRCh38
rs775864895 Chr.11: 72195393-72195393 on Build GRCh38
rs762186998 Chr.11: 72195568-72195568 on Build GRCh38
rs755627957 Chr.11: 72195416-72195416 on Build GRCh38
rs1057518816 Chr.11: 72195389-72195389 on Build GRCh38
rs772210412 Chr.11: 72195435-72195435 on Build GRCh38
rs764487833 Chr.11: 72195504-72195504 on Build GRCh38
rs771232058 Chr.11: 72195491-72195491 on Build GRCh38
rs1055116928 Chr.11: 72195545-72195545 on Build GRCh38
rs145674759 Chr.11: 72195423-72195423 on Build GRCh38
rs201889701 Chr.11: 72195575-72195575 on Build GRCh38
rs759099123 Chr.11: 72195428-72195429 on Build GRCh38
rs776901356 Chr.11: 72195381-72195381 on Build GRCh38
rs767252235 Chr.11: 72195430-72195431 on Build GRCh38
rs771104939 Chr.11: 72195376-72195376 on Build GRCh38
rs886042924 Chr.11: 72195467-72195467 on Build GRCh38

View Full Product Details