Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4955755-4956014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO3
Entrez Gene ID
2027
Gene Name
enolase 3
Gene Aliases
GSD13, MSE
UniGene
Hs.224171
Gene Chromosome Location
Chr.17: 4949182-4957131 on Build GRCh38
Transcripts
NM_001193503.1 NM_001976.4 NM_053013.3 XM_005256521.2 XM_011523729.1 XM_017024346.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3519104 c.928G>A p.G310R Chr.17: 4956004-4956004 on Build GRCh38
1718738 c.921C>T p.F307F Chr.17: 4955997-4955997 on Build GRCh38
6219261 c.877G>A p.E293K Chr.17: 4955953-4955953 on Build GRCh38
3519103 c.926C>T p.S309L Chr.17: 4956002-4956002 on Build GRCh38
2699337 c.931G>A p.V311M Chr.17: 4956007-4956007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768148104 Chr.17: 4955916-4955917 on Build GRCh38
rs565982583 Chr.17: 4955991-4955991 on Build GRCh38
rs776868176 Chr.17: 4955981-4955981 on Build GRCh38
rs745352059 Chr.17: 4955933-4955933 on Build GRCh38
rs769010775 Chr.17: 4955964-4955964 on Build GRCh38
rs111645512 Chr.17: 4955828-4955828 on Build GRCh38
rs201494026 Chr.17: 4955938-4955938 on Build GRCh38
rs878983498 Chr.17: 4955817-4955839 on Build GRCh38
rs28619016 Chr.17: 4955818-4955818 on Build GRCh38
rs761516947 Chr.17: 4955928-4955929 on Build GRCh38
rs2309347 Chr.17: 4955817-4955817 on Build GRCh38
rs1018783798 Chr.17: 4955993-4955993 on Build GRCh38
rs747544030 Chr.17: 4955820-4955850 on Build GRCh38
rs140408533 Chr.17: 4955916-4955916 on Build GRCh38
rs928125011 Chr.17: 4955848-4955848 on Build GRCh38
rs772062073 Chr.17: 4955984-4955984 on Build GRCh38
rs879179763 Chr.17: 4955883-4955883 on Build GRCh38
rs1047462222 Chr.17: 4955862-4955862 on Build GRCh38
rs386794781 Chr.17: 4955817-4955883 on Build GRCh38
rs199572580 Chr.17: 4955829-4955829 on Build GRCh38
rs778403016 Chr.17: 4955816-4955817 on Build GRCh38
rs889097238 Chr.17: 4955969-4955969 on Build GRCh38
rs959574518 Chr.17: 4955909-4955909 on Build GRCh38
rs110703 Chr.17: 4955850-4955850 on Build GRCh38
rs1008510 Chr.17: 4955872-4955872 on Build GRCh38
rs71362561 Chr.17: 4955850-4955850 on Build GRCh38
rs1036489616 Chr.17: 4955898-4955899 on Build GRCh38
rs150346560 Chr.17: 4955919-4955919 on Build GRCh38
rs146545678 Chr.17: 4955861-4955861 on Build GRCh38
rs940802565 Chr.17: 4955886-4955886 on Build GRCh38
rs780704553 Chr.17: 4955942-4955942 on Build GRCh38
rs911783232 Chr.17: 4955785-4955785 on Build GRCh38
rs771337952 Chr.17: 4955807-4955808 on Build GRCh38
rs746936509 Chr.17: 4955905-4955906 on Build GRCh38
rs71367864 Chr.17: 4955808-4955818 on Build GRCh38
rs5027280 Chr.17: 4955905-4955905 on Build GRCh38
rs1983153 Chr.17: 4955840-4955840 on Build GRCh38
rs117488294 Chr.17: 4955839-4955839 on Build GRCh38
rs80060797 Chr.17: 4955807-4955807 on Build GRCh38
rs5027279 Chr.17: 4955894-4955894 on Build GRCh38
rs1050837703 Chr.17: 4955925-4955925 on Build GRCh38
rs943275560 Chr.17: 4955795-4955795 on Build GRCh38
rs549943315 Chr.17: 4955842-4955842 on Build GRCh38
rs58519280 Chr.17: 4955894-4955904 on Build GRCh38
rs451069 Chr.17: 4955884-4955884 on Build GRCh38
rs968061465 Chr.17: 4955929-4955929 on Build GRCh38
rs916863174 Chr.17: 4955783-4955783 on Build GRCh38
rs748358058 Chr.17: 4955990-4955990 on Build GRCh38
rs796703622 Chr.17: 4955796-4955796 on Build GRCh38
rs984130141 Chr.17: 4955796-4955828 on Build GRCh38
rs759470373 Chr.17: 4955952-4955952 on Build GRCh38
rs376564824 Chr.17: 4955996-4955996 on Build GRCh38
rs796836227 Chr.17: 4955806-4955806 on Build GRCh38
rs978520383 Chr.17: 4955949-4955949 on Build GRCh38

View Full Product Details