Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95889011-95889236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL2
Entrez Gene ID
22936
Gene Name
elongation factor for RNA polymerase II 2
Gene Aliases
-
UniGene
Hs.192221
Gene Chromosome Location
Chr.5: 95885098-95962288 on Build GRCh38
Transcripts
NM_012081.5 XM_006714575.2 XM_017009239.1 XM_017009240.1 XM_017009241.1 XM_017009242.1 XM_017009243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3715281 c.1786G>T p.E596* Chr.5: 95889106-95889106 on Build GRCh38
6421150 c.1788A>C p.E596D Chr.5: 95889104-95889104 on Build GRCh38
6420643 c.1783C>T p.Q595* Chr.5: 95889109-95889109 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746265461 Chr.5: 95889163-95889163 on Build GRCh38
rs749229622 Chr.5: 95889054-95889054 on Build GRCh38
rs769462915 Chr.5: 95889175-95889175 on Build GRCh38
rs566870740 Chr.5: 95889151-95889151 on Build GRCh38
rs750258365 Chr.5: 95889042-95889042 on Build GRCh38
rs777750798 Chr.5: 95889134-95889134 on Build GRCh38
rs763414952 Chr.5: 95889174-95889174 on Build GRCh38
rs755904077 Chr.5: 95889061-95889061 on Build GRCh38
rs771197675 Chr.5: 95889146-95889146 on Build GRCh38
rs762873736 Chr.5: 95889078-95889079 on Build GRCh38
rs770008048 Chr.5: 95889167-95889167 on Build GRCh38
rs200787935 Chr.5: 95889149-95889149 on Build GRCh38
rs767267861 Chr.5: 95889041-95889041 on Build GRCh38
rs544392198 Chr.5: 95889196-95889196 on Build GRCh38
rs747084414 Chr.5: 95889135-95889135 on Build GRCh38
rs753668462 Chr.5: 95889112-95889112 on Build GRCh38
rs775905401 Chr.5: 95889173-95889173 on Build GRCh38
rs754920792 Chr.5: 95889124-95889124 on Build GRCh38
rs12515492 Chr.5: 95889079-95889079 on Build GRCh38
rs112809853 Chr.5: 95889192-95889192 on Build GRCh38

View Full Product Details