Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45712916-45713151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO46
Entrez Gene ID
23403
Gene Name
F-box protein 46
Gene Aliases
20D7-FC4, FBXO34L, Fbx46
UniGene
Hs.128702
Gene Chromosome Location
Chr.19: 45710629-45730893 on Build GRCh38
Transcripts
NM_001080469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6379770 c.521A>G p.E174G Chr.19: 45712975-45712975 on Build GRCh38
4855768 c.485C>T p.S162L Chr.19: 45713011-45713011 on Build GRCh38
5755412 c.368A>G p.H123R Chr.19: 45713128-45713128 on Build GRCh38
998274 c.482C>T p.A161V Chr.19: 45713014-45713014 on Build GRCh38
4817036 c.436G>C p.D146H Chr.19: 45713060-45713060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375341217 Chr.19: 45713030-45713030 on Build GRCh38
rs867003742 Chr.19: 45712979-45712979 on Build GRCh38
rs773570673 Chr.19: 45713015-45713015 on Build GRCh38
rs909709953 Chr.19: 45713067-45713067 on Build GRCh38
rs111806912 Chr.19: 45713018-45713018 on Build GRCh38
rs761662292 Chr.19: 45713096-45713096 on Build GRCh38
rs960893118 Chr.19: 45712996-45712996 on Build GRCh38
rs1006407452 Chr.19: 45712947-45712947 on Build GRCh38
rs771350349 Chr.19: 45713022-45713022 on Build GRCh38
rs369102631 Chr.19: 45713006-45713006 on Build GRCh38
rs997822961 Chr.19: 45712945-45712945 on Build GRCh38
rs942136585 Chr.19: 45713061-45713061 on Build GRCh38
rs974807728 Chr.19: 45713027-45713027 on Build GRCh38
rs746588517 Chr.19: 45712988-45712988 on Build GRCh38
rs748618687 Chr.19: 45713056-45713056 on Build GRCh38
rs374509869 Chr.19: 45713089-45713089 on Build GRCh38
rs552011140 Chr.19: 45712938-45712938 on Build GRCh38
rs762845625 Chr.19: 45713117-45713117 on Build GRCh38
rs767670697 Chr.19: 45713029-45713029 on Build GRCh38
rs941169944 Chr.19: 45713121-45713121 on Build GRCh38
rs766384799 Chr.19: 45712940-45712940 on Build GRCh38
rs534388646 Chr.19: 45713007-45713007 on Build GRCh38
rs1037910375 Chr.19: 45713086-45713086 on Build GRCh38
rs763016254 Chr.19: 45712991-45712991 on Build GRCh38
rs766068911 Chr.19: 45713046-45713046 on Build GRCh38
rs778727139 Chr.19: 45712977-45712977 on Build GRCh38
rs929358066 Chr.19: 45713043-45713043 on Build GRCh38
rs773009792 Chr.19: 45713115-45713115 on Build GRCh38
rs778142091 Chr.19: 45713075-45713075 on Build GRCh38
rs766121787 Chr.19: 45713118-45713118 on Build GRCh38
rs886582064 Chr.19: 45712944-45712944 on Build GRCh38
rs758010575 Chr.19: 45712980-45712980 on Build GRCh38
rs751626785 Chr.19: 45712943-45712943 on Build GRCh38
rs764180636 Chr.19: 45712952-45712952 on Build GRCh38
rs574259552 Chr.19: 45713041-45713041 on Build GRCh38
rs759735318 Chr.19: 45713025-45713025 on Build GRCh38
rs764953697 Chr.19: 45713039-45713039 on Build GRCh38
rs924986517 Chr.19: 45713019-45713019 on Build GRCh38
rs770079940 Chr.19: 45713013-45713013 on Build GRCh38
rs749723906 Chr.19: 45713076-45713076 on Build GRCh38
rs756580139 Chr.19: 45713069-45713069 on Build GRCh38
rs148986286 Chr.19: 45712998-45712998 on Build GRCh38
rs774706193 Chr.19: 45713024-45713024 on Build GRCh38
rs905947726 Chr.19: 45712953-45712953 on Build GRCh38
rs753800301 Chr.19: 45712968-45712968 on Build GRCh38
rs765286089 Chr.19: 45713113-45713113 on Build GRCh38
rs779543785 Chr.19: 45712982-45712982 on Build GRCh38
rs750164936 Chr.19: 45713040-45713040 on Build GRCh38
rs374032371 Chr.19: 45713047-45713047 on Build GRCh38
rs775580429 Chr.19: 45713095-45713095 on Build GRCh38
rs371124111 Chr.19: 45713087-45713087 on Build GRCh38
rs771395557 Chr.19: 45713079-45713079 on Build GRCh38
rs759689203 Chr.19: 45712950-45712950 on Build GRCh38
rs780634187 Chr.19: 45713053-45713053 on Build GRCh38
rs372228121 Chr.19: 45712976-45712976 on Build GRCh38
rs772298721 Chr.19: 45713090-45713090 on Build GRCh38
rs369724742 Chr.19: 45713036-45713036 on Build GRCh38
rs377664802 Chr.19: 45713048-45713048 on Build GRCh38

View Full Product Details