Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22613957-22614222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCAR2
Entrez Gene ID
57805
Gene Name
cell cycle and apoptosis regulator 2
Gene Aliases
DBC-1, DBC1, KIAA1967, NET35, p30 DBC, p30DBC
UniGene
Hs.744848
Gene Chromosome Location
Chr.8: 22604632-22620471 on Build GRCh38
Transcripts
NM_021174.5 NR_033902.1 XM_011544603.1 XM_011544604.1 XM_017013717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3731006 c.815C>A p.P272H Chr.8: 22614202-22614202 on Build GRCh38
3898964 c.799T>G p.F267V Chr.8: 22614186-22614186 on Build GRCh38
3732773 c.823C>T p.L275F Chr.8: 22614210-22614210 on Build GRCh38
7293589 c.800T>C p.F267S Chr.8: 22614187-22614187 on Build GRCh38
6201736 c.784A>C p.S262R Chr.8: 22614171-22614171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755655299 Chr.8: 22614123-22614123 on Build GRCh38
rs921998950 Chr.8: 22614020-22614020 on Build GRCh38
rs556307922 Chr.8: 22614097-22614097 on Build GRCh38
rs374484328 Chr.8: 22614190-22614190 on Build GRCh38
rs199676303 Chr.8: 22614058-22614058 on Build GRCh38
rs138410466 Chr.8: 22614063-22614063 on Build GRCh38
rs200842766 Chr.8: 22614042-22614042 on Build GRCh38
rs202069177 Chr.8: 22614044-22614044 on Build GRCh38
rs772238002 Chr.8: 22614156-22614156 on Build GRCh38
rs752126428 Chr.8: 22614118-22614118 on Build GRCh38
rs752109355 Chr.8: 22614122-22614122 on Build GRCh38
rs200102554 Chr.8: 22614069-22614069 on Build GRCh38
rs7357458 Chr.8: 22614185-22614185 on Build GRCh38
rs199873807 Chr.8: 22614158-22614158 on Build GRCh38
rs757965399 Chr.8: 22614139-22614139 on Build GRCh38
rs202239293 Chr.8: 22614161-22614161 on Build GRCh38
rs534556020 Chr.8: 22614029-22614029 on Build GRCh38
rs777515709 Chr.8: 22614130-22614130 on Build GRCh38
rs767011807 Chr.8: 22614192-22614192 on Build GRCh38
rs762417033 Chr.8: 22614030-22614030 on Build GRCh38
rs117666963 Chr.8: 22614007-22614007 on Build GRCh38
rs776997162 Chr.8: 22614170-22614170 on Build GRCh38
rs901639616 Chr.8: 22614023-22614023 on Build GRCh38
rs768836652 Chr.8: 22614168-22614168 on Build GRCh38
rs201034717 Chr.8: 22614096-22614096 on Build GRCh38
rs781014824 Chr.8: 22614114-22614114 on Build GRCh38
rs200838587 Chr.8: 22614060-22614060 on Build GRCh38
rs202200459 Chr.8: 22614188-22614188 on Build GRCh38
rs1021274676 Chr.8: 22614088-22614088 on Build GRCh38
rs754682907 Chr.8: 22614184-22614185 on Build GRCh38
rs754726967 Chr.8: 22614105-22614105 on Build GRCh38
rs775380486 Chr.8: 22614059-22614059 on Build GRCh38
rs915979172 Chr.8: 22614197-22614197 on Build GRCh38
rs558989325 Chr.8: 22614057-22614057 on Build GRCh38
rs147153683 Chr.8: 22614189-22614189 on Build GRCh38
rs200966293 Chr.8: 22614193-22614193 on Build GRCh38
rs200096133 Chr.8: 22614162-22614162 on Build GRCh38
rs1043576329 Chr.8: 22614031-22614031 on Build GRCh38
rs773234896 Chr.8: 22614046-22614046 on Build GRCh38
rs749308771 Chr.8: 22614135-22614135 on Build GRCh38
rs200987523 Chr.8: 22614160-22614160 on Build GRCh38
rs770729470 Chr.8: 22614051-22614051 on Build GRCh38
rs201209039 Chr.8: 22614176-22614176 on Build GRCh38
rs764599258 Chr.8: 22614041-22614041 on Build GRCh38
rs748970227 Chr.8: 22614049-22614049 on Build GRCh38
rs754336801 Chr.8: 22614008-22614008 on Build GRCh38
rs764290754 Chr.8: 22613982-22613982 on Build GRCh38
rs373792843 Chr.8: 22614061-22614061 on Build GRCh38
rs976481992 Chr.8: 22614200-22614200 on Build GRCh38
rs753484766 Chr.8: 22614082-22614083 on Build GRCh38
rs940620452 Chr.8: 22614001-22614001 on Build GRCh38
rs116637032 Chr.8: 22614098-22614098 on Build GRCh38
rs774545278 Chr.8: 22614056-22614056 on Build GRCh38
rs368910038 Chr.8: 22614150-22614150 on Build GRCh38
rs778724078 Chr.8: 22614138-22614138 on Build GRCh38
rs770711234 Chr.8: 22614137-22614137 on Build GRCh38

View Full Product Details