Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6217455-6217682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf42
Entrez Gene ID
160298
Gene Name
chromosome 11 open reading frame 42
Gene Aliases
-
UniGene
Hs.278221
Gene Chromosome Location
Chr.11: 6202997-6226783 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Gene Symbol
FAM160A2
Entrez Gene ID
84067
Gene Name
family with sequence similarity 160 member A2
Gene Aliases
C11orf56
UniGene
Hs.501793
Gene Chromosome Location
Chr.11: 6211334-6234711 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5900776 c.2105G>T p.G702V Chr.11: 6217523-6217523 on Build GRCh38
4682644 c.2093C>T p.A698V Chr.11: 6217535-6217535 on Build GRCh38
5880361 c.2140_2141insG p.L714fs*4 Chr.11: 6217487-6217488 on Build GRCh38
7080820 c.2065G>C p.G689R Chr.11: 6217563-6217563 on Build GRCh38
4433622 c.1986G>C p.K662N Chr.11: 6217642-6217642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17852844 Chr.11: 6217635-6217635 on Build GRCh38
rs568608846 Chr.11: 6217547-6217547 on Build GRCh38
rs764741733 Chr.11: 6217521-6217521 on Build GRCh38
rs757453187 Chr.11: 6217641-6217641 on Build GRCh38
rs946017205 Chr.11: 6217531-6217531 on Build GRCh38
rs367551860 Chr.11: 6217530-6217530 on Build GRCh38
rs777911426 Chr.11: 6217637-6217637 on Build GRCh38
rs145525932 Chr.11: 6217614-6217614 on Build GRCh38
rs77795471 Chr.11: 6217626-6217626 on Build GRCh38
rs774428676 Chr.11: 6217505-6217505 on Build GRCh38
rs762502154 Chr.11: 6217606-6217606 on Build GRCh38
rs200320410 Chr.11: 6217648-6217648 on Build GRCh38
rs114090040 Chr.11: 6217540-6217540 on Build GRCh38
rs775660803 Chr.11: 6217595-6217595 on Build GRCh38
rs907341260 Chr.11: 6217534-6217534 on Build GRCh38
rs149699306 Chr.11: 6217553-6217553 on Build GRCh38
rs990279801 Chr.11: 6217659-6217659 on Build GRCh38
rs781243507 Chr.11: 6217477-6217477 on Build GRCh38
rs772120174 Chr.11: 6217660-6217660 on Build GRCh38
rs749441839 Chr.11: 6217582-6217582 on Build GRCh38
rs146377412 Chr.11: 6217597-6217597 on Build GRCh38
rs780876381 Chr.11: 6217541-6217541 on Build GRCh38
rs745909587 Chr.11: 6217620-6217622 on Build GRCh38
rs374838753 Chr.11: 6217514-6217514 on Build GRCh38
rs1025768459 Chr.11: 6217611-6217611 on Build GRCh38
rs769793886 Chr.11: 6217561-6217561 on Build GRCh38
rs746123727 Chr.11: 6217653-6217653 on Build GRCh38
rs770028848 Chr.11: 6217493-6217493 on Build GRCh38
rs761129829 Chr.11: 6217520-6217520 on Build GRCh38
rs142130040 Chr.11: 6217590-6217590 on Build GRCh38
rs892053817 Chr.11: 6217578-6217578 on Build GRCh38
rs376254533 Chr.11: 6217575-6217575 on Build GRCh38
rs964156157 Chr.11: 6217651-6217651 on Build GRCh38
rs771117512 Chr.11: 6217584-6217584 on Build GRCh38
rs1049096279 Chr.11: 6217478-6217478 on Build GRCh38
rs369189662 Chr.11: 6217508-6217508 on Build GRCh38
rs749214656 Chr.11: 6217638-6217638 on Build GRCh38
rs371804376 Chr.11: 6217511-6217511 on Build GRCh38
rs752470746 Chr.11: 6217632-6217632 on Build GRCh38
rs754331058 Chr.11: 6217605-6217605 on Build GRCh38
rs773373606 Chr.11: 6217500-6217500 on Build GRCh38
rs771207253 Chr.11: 6217504-6217504 on Build GRCh38
rs201173178 Chr.11: 6217591-6217591 on Build GRCh38
rs780812920 Chr.11: 6217551-6217551 on Build GRCh38
rs141597566 Chr.11: 6217484-6217484 on Build GRCh38
rs751259355 Chr.11: 6217616-6217616 on Build GRCh38
rs200204858 Chr.11: 6217550-6217550 on Build GRCh38
rs774682640 Chr.11: 6217585-6217585 on Build GRCh38
rs866799938 Chr.11: 6217580-6217580 on Build GRCh38
rs764652326 Chr.11: 6217604-6217604 on Build GRCh38
rs763278437 Chr.11: 6217503-6217503 on Build GRCh38

View Full Product Details