Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 139457698-139457935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECSCR
Entrez Gene ID
641700
Gene Name
endothelial cell surface expressed chemotaxis and apoptosis regulator
Gene Aliases
ARIA, ECSM2
UniGene
Hs.483538
Gene Chromosome Location
Chr.5: 139448554-139462743 on Build GRCh38
Transcripts
NM_001077693.3 NM_001293739.1 NR_121659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4448054 c.128T>A p.L43Q Chr.5: 139457786-139457786 on Build GRCh38
5402231 c.154C>T p.P52S Chr.5: 139457760-139457760 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1007108773 Chr.5: 139457889-139457889 on Build GRCh38
rs758432814 Chr.5: 139457821-139457821 on Build GRCh38
rs376230659 Chr.5: 139457842-139457842 on Build GRCh38
rs1024419719 Chr.5: 139457760-139457760 on Build GRCh38
rs763863068 Chr.5: 139457799-139457799 on Build GRCh38
rs1049386672 Chr.5: 139457896-139457896 on Build GRCh38
rs576391767 Chr.5: 139457775-139457775 on Build GRCh38
rs540687630 Chr.5: 139457881-139457881 on Build GRCh38
rs755152752 Chr.5: 139457841-139457841 on Build GRCh38
rs369292719 Chr.5: 139457846-139457846 on Build GRCh38
rs887673454 Chr.5: 139457906-139457906 on Build GRCh38
rs910232288 Chr.5: 139457829-139457829 on Build GRCh38
rs780122088 Chr.5: 139457833-139457833 on Build GRCh38
rs375731899 Chr.5: 139457754-139457754 on Build GRCh38
rs753569039 Chr.5: 139457803-139457803 on Build GRCh38
rs776847677 Chr.5: 139457741-139457741 on Build GRCh38
rs147720581 Chr.5: 139457797-139457797 on Build GRCh38
rs1018473764 Chr.5: 139457894-139457894 on Build GRCh38
rs773907058 Chr.5: 139457776-139457776 on Build GRCh38
rs748442607 Chr.5: 139457845-139457845 on Build GRCh38
rs967485044 Chr.5: 139457790-139457790 on Build GRCh38
rs565065683 Chr.5: 139457819-139457819 on Build GRCh38
rs1000366322 Chr.5: 139457870-139457870 on Build GRCh38
rs759071078 Chr.5: 139457792-139457792 on Build GRCh38
rs765135807 Chr.5: 139457816-139457816 on Build GRCh38
rs1002560955 Chr.5: 139457913-139457913 on Build GRCh38
rs767171879 Chr.5: 139457795-139457795 on Build GRCh38
rs929086666 Chr.5: 139457873-139457873 on Build GRCh38
rs747172897 Chr.5: 139457835-139457835 on Build GRCh38
rs944438583 Chr.5: 139457834-139457834 on Build GRCh38
rs201654370 Chr.5: 139457796-139457796 on Build GRCh38

View Full Product Details