Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32273051-32273324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPC1
Entrez Gene ID
80314
Gene Name
enhancer of polycomb homolog 1
Gene Aliases
Epl1
UniGene
Hs.167805
Gene Chromosome Location
Chr.10: 32267716-32378798 on Build GRCh38
Transcripts
NM_001272004.1 NM_001272019.2 NM_001282391.1 NM_025209.3 XM_006717510.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7163470 c.1810G>A p.A604T Chr.10: 32273216-32273216 on Build GRCh38
4964310 c.1756G>A p.E586K Chr.10: 32273270-32273270 on Build GRCh38
5038869 c.1863+1G>T p.? Chr.10: 32273162-32273162 on Build GRCh38
5698158 c.1759C>G p.Q587E Chr.10: 32273267-32273267 on Build GRCh38
2157529 c.1756G>T p.E586* Chr.10: 32273270-32273270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898323264 Chr.10: 32273226-32273226 on Build GRCh38
rs772626611 Chr.10: 32273156-32273156 on Build GRCh38
rs753690577 Chr.10: 32273238-32273238 on Build GRCh38
rs775923864 Chr.10: 32273169-32273169 on Build GRCh38
rs539774588 Chr.10: 32273265-32273265 on Build GRCh38
rs750409783 Chr.10: 32273284-32273284 on Build GRCh38
rs944702321 Chr.10: 32273155-32273155 on Build GRCh38
rs776962013 Chr.10: 32273179-32273179 on Build GRCh38
rs200969067 Chr.10: 32273139-32273139 on Build GRCh38
rs907955208 Chr.10: 32273148-32273148 on Build GRCh38
rs769225263 Chr.10: 32273176-32273176 on Build GRCh38
rs777513875 Chr.10: 32273095-32273095 on Build GRCh38
rs779320532 Chr.10: 32273141-32273141 on Build GRCh38
rs535300277 Chr.10: 32273180-32273180 on Build GRCh38
rs752551109 Chr.10: 32273233-32273233 on Build GRCh38
rs755616855 Chr.10: 32273295-32273298 on Build GRCh38
rs756081923 Chr.10: 32273089-32273089 on Build GRCh38
rs557162371 Chr.10: 32273189-32273189 on Build GRCh38
rs150143347 Chr.10: 32273216-32273216 on Build GRCh38
rs749410014 Chr.10: 32273127-32273127 on Build GRCh38
rs930723320 Chr.10: 32273133-32273133 on Build GRCh38
rs746069118 Chr.10: 32273153-32273153 on Build GRCh38
rs746163827 Chr.10: 32273272-32273272 on Build GRCh38
rs111410751 Chr.10: 32273243-32273243 on Build GRCh38
rs141719918 Chr.10: 32273211-32273211 on Build GRCh38
rs568202307 Chr.10: 32273173-32273173 on Build GRCh38
rs919801378 Chr.10: 32273125-32273125 on Build GRCh38
rs576438106 Chr.10: 32273213-32273213 on Build GRCh38
rs759184073 Chr.10: 32273214-32273214 on Build GRCh38
rs772432767 Chr.10: 32273285-32273285 on Build GRCh38
rs765851603 Chr.10: 32273181-32273181 on Build GRCh38
rs755817235 Chr.10: 32273237-32273237 on Build GRCh38
rs188745406 Chr.10: 32273202-32273202 on Build GRCh38
rs757463750 Chr.10: 32273250-32273250 on Build GRCh38
rs767542037 Chr.10: 32273183-32273184 on Build GRCh38

View Full Product Details