Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49804885-49805146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM212A
Entrez Gene ID
389119
Gene Name
family with sequence similarity 212 member A
Gene Aliases
C3orf54, INKA1
UniGene
Hs.86674
Gene Chromosome Location
Chr.3: 49803130-49805030 on Build GRCh38
Transcripts
NM_203370.1 XM_017006372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1309286 c.803C>A p.A268E Chr.3: 49804926-49804926 on Build GRCh38
1238370 c.796C>T p.R266C Chr.3: 49804919-49804919 on Build GRCh38
4158161 c.763C>T p.R255C Chr.3: 49804886-49804886 on Build GRCh38
7099106 c.821G>A p.R274H Chr.3: 49804944-49804944 on Build GRCh38
5703051 c.797G>A p.R266H Chr.3: 49804920-49804920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779030774 Chr.3: 49805032-49805032 on Build GRCh38
rs367793741 Chr.3: 49804982-49804982 on Build GRCh38
rs778061087 Chr.3: 49804907-49804907 on Build GRCh38
rs760182460 Chr.3: 49804963-49804963 on Build GRCh38
rs759272477 Chr.3: 49804952-49804952 on Build GRCh38
rs929710379 Chr.3: 49804960-49804960 on Build GRCh38
rs776374917 Chr.3: 49804944-49804944 on Build GRCh38
rs771067660 Chr.3: 49804909-49804909 on Build GRCh38
rs760471919 Chr.3: 49804938-49804938 on Build GRCh38
rs374620433 Chr.3: 49805029-49805029 on Build GRCh38
rs558825347 Chr.3: 49805050-49805050 on Build GRCh38
rs187938451 Chr.3: 49805088-49805088 on Build GRCh38
rs778691876 Chr.3: 49804915-49804915 on Build GRCh38
rs768123817 Chr.3: 49804943-49804943 on Build GRCh38
rs540891112 Chr.3: 49805069-49805069 on Build GRCh38
rs745791784 Chr.3: 49805034-49805034 on Build GRCh38
rs758270733 Chr.3: 49805036-49805036 on Build GRCh38
rs35210843 Chr.3: 49805054-49805054 on Build GRCh38
rs886627125 Chr.3: 49805021-49805021 on Build GRCh38
rs72938108 Chr.3: 49805082-49805082 on Build GRCh38
rs766223998 Chr.3: 49804936-49804936 on Build GRCh38
rs768155766 Chr.3: 49804968-49804968 on Build GRCh38
rs749289018 Chr.3: 49804908-49804908 on Build GRCh38
rs942371771 Chr.3: 49804920-49804920 on Build GRCh38
rs754403394 Chr.3: 49805101-49805101 on Build GRCh38
rs528521118 Chr.3: 49805046-49805046 on Build GRCh38
rs147770013 Chr.3: 49804906-49804906 on Build GRCh38
rs1030334166 Chr.3: 49805123-49805123 on Build GRCh38
rs756534866 Chr.3: 49804993-49804993 on Build GRCh38
rs149900702 Chr.3: 49804950-49804950 on Build GRCh38
rs775322243 Chr.3: 49804926-49804926 on Build GRCh38
rs370093341 Chr.3: 49805022-49805022 on Build GRCh38
rs771849514 Chr.3: 49804925-49804925 on Build GRCh38
rs745844777 Chr.3: 49804919-49804919 on Build GRCh38
rs764845760 Chr.3: 49804949-49804949 on Build GRCh38
rs766261212 Chr.3: 49804941-49804943 on Build GRCh38
rs1001670157 Chr.3: 49805049-49805049 on Build GRCh38
rs368644051 Chr.3: 49805030-49805030 on Build GRCh38
rs967390141 Chr.3: 49805091-49805091 on Build GRCh38

View Full Product Details