Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157766790-157767035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL2
Entrez Gene ID
79368
Gene Name
Fc receptor like 2
Gene Aliases
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C
UniGene
Hs.437393
Gene Chromosome Location
Chr.1: 157745733-157777472 on Build GRCh38
Transcripts
NM_001159488.1 NM_030764.3 NR_125358.1 XM_011509974.2 XM_011509975.2 XM_011509976.2 XM_017002316.1 XM_017002317.1 XM_017002318.1 XM_017002319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3418169 c.1183G>T p.D395Y Chr.1: 157766951-157766951 on Build GRCh38
6308915 c.1252T>A p.L418M Chr.1: 157766882-157766882 on Build GRCh38
6662873 c.493T>C p.L165L Chr.1: 157766882-157766882 on Build GRCh38
3475820 c.515T>A p.I172K Chr.1: 157766860-157766860 on Build GRCh38
3475821 c.1211G>A p.G404E Chr.1: 157766923-157766923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766822397 Chr.1: 157766908-157766908 on Build GRCh38
rs778672969 Chr.1: 157766974-157766974 on Build GRCh38
rs750460981 Chr.1: 157766842-157766842 on Build GRCh38
rs748994970 Chr.1: 157766829-157766829 on Build GRCh38
rs954314638 Chr.1: 157766862-157766862 on Build GRCh38
rs199754690 Chr.1: 157766970-157766970 on Build GRCh38
rs767149698 Chr.1: 157766941-157766941 on Build GRCh38
rs202079887 Chr.1: 157766859-157766859 on Build GRCh38
rs141224256 Chr.1: 157766884-157766884 on Build GRCh38
rs1019788204 Chr.1: 157766855-157766855 on Build GRCh38
rs751325876 Chr.1: 157766910-157766910 on Build GRCh38
rs1031282506 Chr.1: 157766922-157766922 on Build GRCh38
rs138049282 Chr.1: 157766893-157766893 on Build GRCh38
rs747198894 Chr.1: 157766861-157766861 on Build GRCh38
rs755131853 Chr.1: 157766998-157766998 on Build GRCh38
rs34445670 Chr.1: 157766883-157766883 on Build GRCh38
rs1037053898 Chr.1: 157766833-157766833 on Build GRCh38
rs768787443 Chr.1: 157766867-157766867 on Build GRCh38
rs772359785 Chr.1: 157766851-157766851 on Build GRCh38
rs978335006 Chr.1: 157766909-157766909 on Build GRCh38
rs756107904 Chr.1: 157766956-157766956 on Build GRCh38
rs777628519 Chr.1: 157766822-157766822 on Build GRCh38
rs371790323 Chr.1: 157767003-157767003 on Build GRCh38
rs149515945 Chr.1: 157766958-157766958 on Build GRCh38
rs753790921 Chr.1: 157766964-157766964 on Build GRCh38
rs776675210 Chr.1: 157766872-157766872 on Build GRCh38
rs995578543 Chr.1: 157766854-157766854 on Build GRCh38
rs952990731 Chr.1: 157766945-157766945 on Build GRCh38
rs1007066276 Chr.1: 157766870-157766870 on Build GRCh38
rs762391165 Chr.1: 157766875-157766875 on Build GRCh38
rs908771296 Chr.1: 157766979-157766979 on Build GRCh38
rs759250625 Chr.1: 157766914-157766914 on Build GRCh38
rs376016153 Chr.1: 157766953-157766953 on Build GRCh38
rs879078882 Chr.1: 157766989-157766989 on Build GRCh38
rs755193542 Chr.1: 157766996-157766996 on Build GRCh38
rs567511739 Chr.1: 157767000-157767000 on Build GRCh38
rs368388157 Chr.1: 157766986-157766986 on Build GRCh38
rs766898179 Chr.1: 157766997-157766998 on Build GRCh38
rs770164116 Chr.1: 157766876-157766876 on Build GRCh38

View Full Product Details