Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81283764-81284037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FABP5
Entrez Gene ID
2171
Gene Name
fatty acid binding protein 5
Gene Aliases
E-FABP, EFABP, KFABP, PA-FABP, PAFABP
UniGene
Hs.408061
Gene Chromosome Location
Chr.8: 81280483-81284777 on Build GRCh38
Transcripts
NM_001444.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6848871 c.260G>A p.C87Y Chr.8: 81283880-81283880 on Build GRCh38
1101971 c.346T>A p.L116I Chr.8: 81283966-81283966 on Build GRCh38
1101970 c.263A>G p.N88S Chr.8: 81283883-81283883 on Build GRCh38
3651274 c.304G>A p.G102R Chr.8: 81283924-81283924 on Build GRCh38
6324611 c.261C>T p.C87C Chr.8: 81283881-81283881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182371778 Chr.8: 81283821-81283821 on Build GRCh38
rs142936320 Chr.8: 81283920-81283920 on Build GRCh38
rs745885500 Chr.8: 81283968-81283968 on Build GRCh38
rs771704545 Chr.8: 81283857-81283857 on Build GRCh38
rs431905490 Chr.8: 81283951-81283951 on Build GRCh38
rs775408791 Chr.8: 81284005-81284007 on Build GRCh38
rs749917972 Chr.8: 81283940-81283940 on Build GRCh38
rs775743786 Chr.8: 81283994-81283994 on Build GRCh38
rs774544819 Chr.8: 81283892-81283892 on Build GRCh38
rs749705596 Chr.8: 81283959-81283959 on Build GRCh38
rs770651593 Chr.8: 81283874-81283877 on Build GRCh38
rs753714375 Chr.8: 81283930-81283930 on Build GRCh38
rs753725509 Chr.8: 81283838-81283838 on Build GRCh38
rs760784784 Chr.8: 81283998-81283998 on Build GRCh38
rs150855563 Chr.8: 81283909-81283909 on Build GRCh38
rs756740644 Chr.8: 81283842-81283842 on Build GRCh38
rs146288109 Chr.8: 81283837-81283837 on Build GRCh38
rs199758626 Chr.8: 81283894-81283894 on Build GRCh38
rs889947968 Chr.8: 81283964-81283964 on Build GRCh38
rs987032466 Chr.8: 81283881-81283881 on Build GRCh38
rs368489863 Chr.8: 81283946-81283946 on Build GRCh38
rs75108814 Chr.8: 81283960-81283960 on Build GRCh38
rs1004928546 Chr.8: 81283825-81283825 on Build GRCh38
rs771618416 Chr.8: 81283889-81283889 on Build GRCh38
rs745619610 Chr.8: 81283975-81283976 on Build GRCh38
rs773732171 Chr.8: 81283882-81283882 on Build GRCh38
rs770055478 Chr.8: 81283876-81283876 on Build GRCh38
rs763394711 Chr.8: 81283883-81283883 on Build GRCh38
rs757793202 Chr.8: 81283942-81283942 on Build GRCh38
rs769680752 Chr.8: 81283997-81284000 on Build GRCh38
rs776447932 Chr.8: 81283944-81283946 on Build GRCh38
rs960674531 Chr.8: 81283850-81283850 on Build GRCh38
rs431905489 Chr.8: 81283899-81283899 on Build GRCh38
rs759757795 Chr.8: 81283897-81283897 on Build GRCh38
rs763815393 Chr.8: 81283921-81283921 on Build GRCh38
rs777710390 Chr.8: 81283834-81283834 on Build GRCh38
rs201830389 Chr.8: 81283888-81283888 on Build GRCh38
rs148299679 Chr.8: 81283854-81283854 on Build GRCh38
rs772284450 Chr.8: 81283969-81283969 on Build GRCh38
rs201120689 Chr.8: 81283957-81283957 on Build GRCh38
rs779611485 Chr.8: 81283944-81283944 on Build GRCh38
rs376469447 Chr.8: 81283963-81283963 on Build GRCh38
rs371106428 Chr.8: 81283853-81283853 on Build GRCh38
rs755921591 Chr.8: 81283830-81283830 on Build GRCh38
rs139325159 Chr.8: 81283915-81283915 on Build GRCh38
rs765225550 Chr.8: 81283939-81283939 on Build GRCh38
rs200065410 Chr.8: 81283903-81283903 on Build GRCh38
rs141453643 Chr.8: 81283877-81283877 on Build GRCh38
rs748484774 Chr.8: 81283871-81283871 on Build GRCh38
rs758828368 Chr.8: 81283801-81283801 on Build GRCh38
rs757111208 Chr.8: 81283934-81283934 on Build GRCh38

View Full Product Details