Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10556598-10556837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT2
Entrez Gene ID
2651
Gene Name
glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group)
Gene Aliases
CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1
UniGene
Hs.519884
Gene Chromosome Location
Chr.6: 10521283-10629368 on Build GRCh38
Transcripts
NM_001491.2 NM_145649.4 XM_005248997.3 XM_005248999.2 XM_006715052.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1071712 c.376G>A p.V126M Chr.6: 10556799-10556799 on Build GRCh38
3366242 c.367T>G p.C123G Chr.6: 10556790-10556790 on Build GRCh38
4160089 c.300C>A p.V100V Chr.6: 10556723-10556723 on Build GRCh38
3154116 c.208_210delTCT p.S71delS Chr.6: 10556631-10556633 on Build GRCh38
1071711 c.195G>T p.M65I Chr.6: 10556618-10556618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952158896 Chr.6: 10556738-10556738 on Build GRCh38
rs35537333 Chr.6: 10556753-10556753 on Build GRCh38
rs766770393 Chr.6: 10556654-10556654 on Build GRCh38
rs768365388 Chr.6: 10556708-10556708 on Build GRCh38
rs759083656 Chr.6: 10556631-10556633 on Build GRCh38
rs17637756 Chr.6: 10556677-10556677 on Build GRCh38
rs761911207 Chr.6: 10556648-10556648 on Build GRCh38
rs200739551 Chr.6: 10556729-10556729 on Build GRCh38
rs749080371 Chr.6: 10556702-10556702 on Build GRCh38
rs766684377 Chr.6: 10556732-10556732 on Build GRCh38
rs754744022 Chr.6: 10556802-10556802 on Build GRCh38
rs202175806 Chr.6: 10556795-10556795 on Build GRCh38
rs779715193 Chr.6: 10556793-10556793 on Build GRCh38
rs148669958 Chr.6: 10556808-10556808 on Build GRCh38
rs749097865 Chr.6: 10556634-10556634 on Build GRCh38
rs530885009 Chr.6: 10556649-10556649 on Build GRCh38
rs111998908 Chr.6: 10556691-10556691 on Build GRCh38
rs569056707 Chr.6: 10556726-10556726 on Build GRCh38
rs993886125 Chr.6: 10556767-10556767 on Build GRCh38
rs753184939 Chr.6: 10556764-10556764 on Build GRCh38
rs755750648 Chr.6: 10556686-10556686 on Build GRCh38
rs2230906 Chr.6: 10556639-10556639 on Build GRCh38
rs745450757 Chr.6: 10556682-10556682 on Build GRCh38
rs369944966 Chr.6: 10556633-10556633 on Build GRCh38
rs768618071 Chr.6: 10556635-10556635 on Build GRCh38
rs139460142 Chr.6: 10556673-10556673 on Build GRCh38
rs111305085 Chr.6: 10556631-10556631 on Build GRCh38
rs765537820 Chr.6: 10556762-10556762 on Build GRCh38
rs759899795 Chr.6: 10556759-10556759 on Build GRCh38
rs767716288 Chr.6: 10556721-10556721 on Build GRCh38
rs367734006 Chr.6: 10556678-10556678 on Build GRCh38
rs748020006 Chr.6: 10556811-10556811 on Build GRCh38
rs754215089 Chr.6: 10556667-10556667 on Build GRCh38
rs779623226 Chr.6: 10556700-10556700 on Build GRCh38
rs769512908 Chr.6: 10556629-10556629 on Build GRCh38
rs749914235 Chr.6: 10556780-10556780 on Build GRCh38
rs546942401 Chr.6: 10556772-10556772 on Build GRCh38
rs552308089 Chr.6: 10556651-10556651 on Build GRCh38
rs374331731 Chr.6: 10556674-10556674 on Build GRCh38
rs774294620 Chr.6: 10556709-10556709 on Build GRCh38
rs373471539 Chr.6: 10556800-10556800 on Build GRCh38
rs1040029439 Chr.6: 10556797-10556797 on Build GRCh38
rs772210999 Chr.6: 10556722-10556722 on Build GRCh38
rs758903818 Chr.6: 10556770-10556770 on Build GRCh38
rs890733070 Chr.6: 10556666-10556666 on Build GRCh38

View Full Product Details