Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65579292-65579517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHBP1L1
Entrez Gene ID
254102
Gene Name
EH domain binding protein 1 like 1
Gene Aliases
-
UniGene
Hs.502867
Gene Chromosome Location
Chr.11: 65576034-65592650 on Build GRCh38
Transcripts
NM_001099409.1 XM_005273867.3 XM_005273868.3 XM_005273869.3 XM_005273870.3 XM_006718486.3 XM_006718487.3 XM_017017467.1 XM_017017468.1 XM_017017469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7365985 c.164C>T p.A55V Chr.11: 65579342-65579342 on Build GRCh38
467232 c.223C>T p.P75S Chr.11: 65579401-65579401 on Build GRCh38
4680448 c.212T>C p.V71A Chr.11: 65579390-65579390 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369857321 Chr.11: 65579427-65579427 on Build GRCh38
rs554014179 Chr.11: 65579342-65579342 on Build GRCh38
rs200668380 Chr.11: 65579382-65579382 on Build GRCh38
rs765970705 Chr.11: 65579438-65579438 on Build GRCh38
rs1038859235 Chr.11: 65579428-65579428 on Build GRCh38
rs758940258 Chr.11: 65579471-65579471 on Build GRCh38
rs769231487 Chr.11: 65579378-65579378 on Build GRCh38
rs750135253 Chr.11: 65579366-65579366 on Build GRCh38
rs369211325 Chr.11: 65579445-65579445 on Build GRCh38
rs913728060 Chr.11: 65579468-65579468 on Build GRCh38
rs966954723 Chr.11: 65579345-65579345 on Build GRCh38
rs1046110835 Chr.11: 65579491-65579491 on Build GRCh38
rs900628662 Chr.11: 65579439-65579439 on Build GRCh38
rs201543472 Chr.11: 65579375-65579375 on Build GRCh38
rs761973330 Chr.11: 65579317-65579317 on Build GRCh38
rs367950334 Chr.11: 65579357-65579357 on Build GRCh38
rs762616228 Chr.11: 65579405-65579405 on Build GRCh38
rs779217760 Chr.11: 65579380-65579380 on Build GRCh38
rs372226517 Chr.11: 65579328-65579328 on Build GRCh38
rs774099479 Chr.11: 65579412-65579412 on Build GRCh38
rs1022317148 Chr.11: 65579492-65579492 on Build GRCh38
rs779319049 Chr.11: 65579456-65579456 on Build GRCh38
rs778046940 Chr.11: 65579484-65579484 on Build GRCh38
rs771254529 Chr.11: 65579391-65579391 on Build GRCh38
rs189053067 Chr.11: 65579415-65579415 on Build GRCh38
rs373018527 Chr.11: 65579432-65579432 on Build GRCh38
rs755236331 Chr.11: 65579452-65579452 on Build GRCh38
rs377366932 Chr.11: 65579433-65579433 on Build GRCh38
rs775446715 Chr.11: 65579396-65579396 on Build GRCh38
rs760963466 Chr.11: 65579320-65579320 on Build GRCh38
rs778172782 Chr.11: 65579344-65579344 on Build GRCh38
rs780838500 Chr.11: 65579360-65579360 on Build GRCh38
rs185071236 Chr.11: 65579385-65579385 on Build GRCh38
rs371437074 Chr.11: 65579386-65579386 on Build GRCh38
rs750550016 Chr.11: 65579466-65579489 on Build GRCh38
rs945165262 Chr.11: 65579481-65579481 on Build GRCh38
rs748667524 Chr.11: 65579457-65579457 on Build GRCh38
rs538328800 Chr.11: 65579496-65579496 on Build GRCh38
rs1010957029 Chr.11: 65579487-65579487 on Build GRCh38
rs747272321 Chr.11: 65579388-65579388 on Build GRCh38
rs79261382 Chr.11: 65579410-65579410 on Build GRCh38
rs375692008 Chr.11: 65579354-65579354 on Build GRCh38
rs964776831 Chr.11: 65579318-65579318 on Build GRCh38
rs562782203 Chr.11: 65579368-65579368 on Build GRCh38
rs749634549 Chr.11: 65579377-65579377 on Build GRCh38
rs529957497 Chr.11: 65579420-65579420 on Build GRCh38
rs752693498 Chr.11: 65579449-65579449 on Build GRCh38
rs376949897 Chr.11: 65579358-65579358 on Build GRCh38
rs781206100 Chr.11: 65579439-65579440 on Build GRCh38
rs569599290 Chr.11: 65579450-65579450 on Build GRCh38

View Full Product Details