Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45670564-45670835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPR
Entrez Gene ID
2696
Gene Name
gastric inhibitory polypeptide receptor
Gene Aliases
PGQTL2
UniGene
Hs.251412
Gene Chromosome Location
Chr.19: 45668187-45683724 on Build GRCh38
Transcripts
NM_000164.3 NM_001308418.1 XM_011526709.2 XM_011526710.2 XM_011526711.2 XM_011526713.2 XM_011526714.2 XM_011526715.2 XM_011526716.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3668298 c.121C>A p.R41R Chr.19: 45670683-45670683 on Build GRCh38
3692879 c.73A>T p.T25S Chr.19: 45670635-45670635 on Build GRCh38
4632221 c.157G>A p.A53T Chr.19: 45670719-45670719 on Build GRCh38
4801125 c.129C>A p.R43R Chr.19: 45670691-45670691 on Build GRCh38
6085320 c.112C>A p.R38S Chr.19: 45670674-45670674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551515236 Chr.19: 45670606-45670606 on Build GRCh38
rs995685418 Chr.19: 45670593-45670593 on Build GRCh38
rs764005735 Chr.19: 45670680-45670680 on Build GRCh38
rs771861504 Chr.19: 45670636-45670636 on Build GRCh38
rs1031532803 Chr.19: 45670663-45670663 on Build GRCh38
rs757133211 Chr.19: 45670625-45670625 on Build GRCh38
rs777091593 Chr.19: 45670594-45670594 on Build GRCh38
rs556224201 Chr.19: 45670643-45670643 on Build GRCh38
rs530198066 Chr.19: 45670803-45670803 on Build GRCh38
rs746491542 Chr.19: 45670703-45670703 on Build GRCh38
rs759654048 Chr.19: 45670679-45670679 on Build GRCh38
rs748623857 Chr.19: 45670741-45670741 on Build GRCh38
rs13306399 Chr.19: 45670699-45670699 on Build GRCh38
rs547550048 Chr.19: 45670774-45670774 on Build GRCh38
rs777330704 Chr.19: 45670658-45670658 on Build GRCh38
rs1038500454 Chr.19: 45670642-45670642 on Build GRCh38
rs1024907382 Chr.19: 45670611-45670611 on Build GRCh38
rs565088603 Chr.19: 45670604-45670605 on Build GRCh38
rs755998511 Chr.19: 45670609-45670609 on Build GRCh38
rs758081370 Chr.19: 45670690-45670690 on Build GRCh38
rs745457046 Chr.19: 45670634-45670634 on Build GRCh38
rs779944737 Chr.19: 45670603-45670603 on Build GRCh38
rs752730761 Chr.19: 45670784-45670784 on Build GRCh38
rs918836214 Chr.19: 45670817-45670817 on Build GRCh38
rs149510000 Chr.19: 45670671-45670671 on Build GRCh38
rs779659417 Chr.19: 45670645-45670645 on Build GRCh38
rs200489477 Chr.19: 45670587-45670587 on Build GRCh38
rs768343561 Chr.19: 45670714-45670714 on Build GRCh38
rs773456016 Chr.19: 45670669-45670669 on Build GRCh38
rs746544644 Chr.19: 45670647-45670647 on Build GRCh38
rs773578742 Chr.19: 45670760-45670760 on Build GRCh38
rs778426085 Chr.19: 45670626-45670626 on Build GRCh38
rs769250990 Chr.19: 45670654-45670654 on Build GRCh38
rs544957328 Chr.19: 45670605-45670605 on Build GRCh38
rs555716721 Chr.19: 45670605-45670605 on Build GRCh38
rs781550091 Chr.19: 45670727-45670727 on Build GRCh38
rs771111683 Chr.19: 45670771-45670771 on Build GRCh38
rs527824031 Chr.19: 45670746-45670746 on Build GRCh38
rs146012706 Chr.19: 45670684-45670684 on Build GRCh38
rs777719484 Chr.19: 45670610-45670610 on Build GRCh38
rs751598933 Chr.19: 45670672-45670672 on Build GRCh38
rs767963853 Chr.19: 45670683-45670683 on Build GRCh38
rs567867210 Chr.19: 45670799-45670799 on Build GRCh38
rs564936619 Chr.19: 45670664-45670664 on Build GRCh38
rs139952137 Chr.19: 45670698-45670698 on Build GRCh38
rs762075272 Chr.19: 45670660-45670660 on Build GRCh38
rs763260665 Chr.19: 45670763-45670763 on Build GRCh38
rs770219187 Chr.19: 45670748-45670748 on Build GRCh38
rs1036735951 Chr.19: 45670813-45670813 on Build GRCh38
rs371179322 Chr.19: 45670783-45670783 on Build GRCh38
rs943664891 Chr.19: 45670812-45670812 on Build GRCh38
rs765358914 Chr.19: 45670599-45670599 on Build GRCh38

View Full Product Details