Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174715473-174715746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRA3
Entrez Gene ID
8001
Gene Name
glycine receptor alpha 3
Gene Aliases
-
UniGene
Hs.413099
Gene Chromosome Location
Chr.4: 174642011-174829568 on Build GRCh38
Transcripts
NM_001042543.2 NM_006529.3 XM_011532267.2 XM_017008630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5926465 c.574+1G>T p.? Chr.4: 174715487-174715487 on Build GRCh38
296764 c.520G>C p.D174H Chr.4: 174715542-174715542 on Build GRCh38
5899456 c.535C>T p.P179S Chr.4: 174715527-174715527 on Build GRCh38
586668 c.540G>T p.M180I Chr.4: 174715522-174715522 on Build GRCh38
5018496 c.497C>T p.T166I Chr.4: 174715565-174715565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768058426 Chr.4: 174715532-174715532 on Build GRCh38
rs112866876 Chr.4: 174715668-174715668 on Build GRCh38
rs867192737 Chr.4: 174715715-174715715 on Build GRCh38
rs370369267 Chr.4: 174715512-174715512 on Build GRCh38
rs765044938 Chr.4: 174715558-174715558 on Build GRCh38
rs376020310 Chr.4: 174715509-174715509 on Build GRCh38
rs756148932 Chr.4: 174715619-174715619 on Build GRCh38
rs755823041 Chr.4: 174715713-174715713 on Build GRCh38
rs775116852 Chr.4: 174715518-174715518 on Build GRCh38
rs1029428955 Chr.4: 174715511-174715511 on Build GRCh38
rs974768669 Chr.4: 174715680-174715680 on Build GRCh38
rs147409488 Chr.4: 174715662-174715662 on Build GRCh38
rs1016226099 Chr.4: 174715628-174715628 on Build GRCh38
rs996584442 Chr.4: 174715510-174715510 on Build GRCh38
rs762858460 Chr.4: 174715578-174715578 on Build GRCh38
rs777628123 Chr.4: 174715633-174715633 on Build GRCh38
rs865790626 Chr.4: 174715527-174715527 on Build GRCh38
rs766365423 Chr.4: 174715582-174715582 on Build GRCh38
rs891274311 Chr.4: 174715557-174715557 on Build GRCh38
rs763582913 Chr.4: 174715617-174715618 on Build GRCh38
rs779166710 Chr.4: 174715502-174715502 on Build GRCh38
rs752766438 Chr.4: 174715603-174715603 on Build GRCh38
rs761394888 Chr.4: 174715554-174715554 on Build GRCh38
rs1013865362 Chr.4: 174715692-174715692 on Build GRCh38
rs746585506 Chr.4: 174715525-174715525 on Build GRCh38
rs1004386684 Chr.4: 174715621-174715621 on Build GRCh38
rs1049181124 Chr.4: 174715546-174715546 on Build GRCh38
rs1002254311 Chr.4: 174715632-174715632 on Build GRCh38
rs868525992 Chr.4: 174715500-174715500 on Build GRCh38
rs773035267 Chr.4: 174715566-174715566 on Build GRCh38
rs776289823 Chr.4: 174715553-174715553 on Build GRCh38
rs775056119 Chr.4: 174715616-174715617 on Build GRCh38
rs549639157 Chr.4: 174715701-174715701 on Build GRCh38

View Full Product Details