Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88620663-88620930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML5
Entrez Gene ID
161436
Gene Name
echinoderm microtubule associated protein like 5
Gene Aliases
EMAP-2
UniGene
Hs.325846
Gene Chromosome Location
Chr.14: 88614213-88792752 on Build GRCh38
Transcripts
NM_183387.2 XM_006720070.3 XM_011536528.2 XM_011536530.2 XM_011536531.2 XM_011536532.2 XM_011536533.2 XM_011536534.2 XM_017021062.1 XM_017021063.1 XM_017021064.1 XM_017021065.1 XM_017021066.1 XM_017021067.1 XM_017021068.1 XM_017021069.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1205271 c.5288A>C p.K1763T Chr.14: 88620841-88620841 on Build GRCh38
5789413 c.5293G>C p.G1765R Chr.14: 88620836-88620836 on Build GRCh38
5985357 c.5094T>A p.H1698Q Chr.14: 88620897-88620897 on Build GRCh38
5139660 c.5291delA p.N1764fs*8 Chr.14: 88620838-88620838 on Build GRCh38
7341261 c.5363T>A p.I1788N Chr.14: 88620766-88620766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762833060 Chr.14: 88620770-88620770 on Build GRCh38
rs370186656 Chr.14: 88620739-88620739 on Build GRCh38
rs780601982 Chr.14: 88620720-88620720 on Build GRCh38
rs770275455 Chr.14: 88620711-88620711 on Build GRCh38
rs772171016 Chr.14: 88620851-88620851 on Build GRCh38
rs769067732 Chr.14: 88620889-88620889 on Build GRCh38
rs761730056 Chr.14: 88620740-88620740 on Build GRCh38
rs375765728 Chr.14: 88620854-88620854 on Build GRCh38
rs375791433 Chr.14: 88620724-88620724 on Build GRCh38
rs756773492 Chr.14: 88620850-88620850 on Build GRCh38
rs748426779 Chr.14: 88620710-88620714 on Build GRCh38
rs755875584 Chr.14: 88620863-88620863 on Build GRCh38
rs532298977 Chr.14: 88620869-88620869 on Build GRCh38
rs779228207 Chr.14: 88620890-88620890 on Build GRCh38
rs773411722 Chr.14: 88620764-88620764 on Build GRCh38
rs907378840 Chr.14: 88620873-88620873 on Build GRCh38
rs899267997 Chr.14: 88620735-88620735 on Build GRCh38
rs867215132 Chr.14: 88620843-88620843 on Build GRCh38
rs763902983 Chr.14: 88620800-88620800 on Build GRCh38
rs569882025 Chr.14: 88620816-88620816 on Build GRCh38
rs530921389 Chr.14: 88620767-88620767 on Build GRCh38
rs772152070 Chr.14: 88620744-88620744 on Build GRCh38
rs770103861 Chr.14: 88620734-88620735 on Build GRCh38
rs760956231 Chr.14: 88620765-88620765 on Build GRCh38
rs750066524 Chr.14: 88620857-88620857 on Build GRCh38
rs993492895 Chr.14: 88620759-88620759 on Build GRCh38
rs1028001743 Chr.14: 88620824-88620825 on Build GRCh38
rs76231557 Chr.14: 88620769-88620769 on Build GRCh38
rs748748462 Chr.14: 88620892-88620892 on Build GRCh38

View Full Product Details