Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123768694-123768919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM91A1
Entrez Gene ID
157769
Gene Name
family with sequence similarity 91 member A1
Gene Aliases
-
UniGene
Hs.459174
Gene Chromosome Location
Chr.8: 123768439-123815452 on Build GRCh38
Transcripts
NM_001317917.1 NM_001317918.1 NM_144963.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1755555 c.8T>G p.I3R Chr.8: 123768710-123768710 on Build GRCh38
6289677 c.1A>G p.M1V Chr.8: 123768703-123768703 on Build GRCh38
1095810 c.59C>T p.A20V Chr.8: 123768761-123768761 on Build GRCh38
5647477 c.66G>C p.V22V Chr.8: 123768768-123768768 on Build GRCh38
1095809 c.17A>T p.E6V Chr.8: 123768719-123768719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372450185 Chr.8: 123768778-123768778 on Build GRCh38
rs950311032 Chr.8: 123768873-123768873 on Build GRCh38
rs933699498 Chr.8: 123768845-123768845 on Build GRCh38
rs755290849 Chr.8: 123768759-123768759 on Build GRCh38
rs781732639 Chr.8: 123768805-123768805 on Build GRCh38
rs1018163961 Chr.8: 123768891-123768891 on Build GRCh38
rs528645461 Chr.8: 123768839-123768839 on Build GRCh38
rs144899068 Chr.8: 123768896-123768896 on Build GRCh38
rs765116609 Chr.8: 123768715-123768717 on Build GRCh38
rs764072361 Chr.8: 123768796-123768796 on Build GRCh38
rs752772689 Chr.8: 123768817-123768817 on Build GRCh38
rs771950097 Chr.8: 123768820-123768820 on Build GRCh38
rs773115077 Chr.8: 123768821-123768821 on Build GRCh38
rs77431769 Chr.8: 123768851-123768851 on Build GRCh38
rs530901202 Chr.8: 123768899-123768899 on Build GRCh38
rs761599272 Chr.8: 123768731-123768731 on Build GRCh38
rs752833313 Chr.8: 123768773-123768773 on Build GRCh38
rs765214014 Chr.8: 123768769-123768769 on Build GRCh38
rs893975438 Chr.8: 123768892-123768892 on Build GRCh38
rs767449243 Chr.8: 123768732-123768732 on Build GRCh38
rs889564336 Chr.8: 123768866-123768866 on Build GRCh38
rs762855024 Chr.8: 123768790-123768790 on Build GRCh38
rs758905229 Chr.8: 123768774-123768774 on Build GRCh38
rs113938868 Chr.8: 123768858-123768858 on Build GRCh38
rs760488117 Chr.8: 123768743-123768743 on Build GRCh38
rs34641714 Chr.8: 123768897-123768897 on Build GRCh38
rs1006652389 Chr.8: 123768867-123768867 on Build GRCh38
rs376499374 Chr.8: 123768779-123768779 on Build GRCh38
rs778221631 Chr.8: 123768819-123768819 on Build GRCh38
rs748880080 Chr.8: 123768806-123768806 on Build GRCh38
rs757672696 Chr.8: 123768797-123768797 on Build GRCh38
rs772955315 Chr.8: 123768739-123768739 on Build GRCh38
rs766214631 Chr.8: 123768752-123768752 on Build GRCh38
rs753980063 Chr.8: 123768758-123768758 on Build GRCh38
rs775356852 Chr.8: 123768778-123768779 on Build GRCh38

View Full Product Details