Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37119880-37120105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2AK2
Entrez Gene ID
5610
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 2
Gene Aliases
EIF2AK1, PKR, PPP1R83, PRKR
UniGene
Hs.131431
Gene Chromosome Location
Chr.2: 37099210-37157065 on Build GRCh38
Transcripts
NM_001135651.2 NM_001135652.2 NM_002759.3 XM_011532987.2 XM_017004503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7236786 c.1159G>C p.D387H Chr.2: 37120048-37120048 on Build GRCh38
6652630 c.1227delA p.K409fs*2 Chr.2: 37119980-37119980 on Build GRCh38
477380 c.1131G>C p.W377C Chr.2: 37120076-37120076 on Build GRCh38
5687813 c.1239A>G p.R413R Chr.2: 37119968-37119968 on Build GRCh38
7199394 c.1228T>A p.L410I Chr.2: 37119979-37119979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761998681 Chr.2: 37120063-37120063 on Build GRCh38
rs750549333 Chr.2: 37119921-37119922 on Build GRCh38
rs764479308 Chr.2: 37120058-37120058 on Build GRCh38
rs769613213 Chr.2: 37120011-37120011 on Build GRCh38
rs780108272 Chr.2: 37119964-37119964 on Build GRCh38
rs750549637 Chr.2: 37119918-37119918 on Build GRCh38
rs765483661 Chr.2: 37119993-37119993 on Build GRCh38
rs778283176 Chr.2: 37120057-37120057 on Build GRCh38
rs748872372 Chr.2: 37120042-37120042 on Build GRCh38
rs979186146 Chr.2: 37119910-37119910 on Build GRCh38
rs757500057 Chr.2: 37119911-37119911 on Build GRCh38
rs754744210 Chr.2: 37119989-37119989 on Build GRCh38
rs35102011 Chr.2: 37119930-37119931 on Build GRCh38
rs1020873428 Chr.2: 37119917-37119917 on Build GRCh38
rs3087381 Chr.2: 37119920-37119921 on Build GRCh38
rs758320164 Chr.2: 37119928-37119928 on Build GRCh38
rs774112406 Chr.2: 37119936-37119939 on Build GRCh38
rs770349755 Chr.2: 37120051-37120051 on Build GRCh38
rs750498734 Chr.2: 37120079-37120079 on Build GRCh38
rs4648217 Chr.2: 37119919-37119920 on Build GRCh38
rs748366481 Chr.2: 37119920-37119923 on Build GRCh38
rs776003747 Chr.2: 37120054-37120054 on Build GRCh38
rs765268481 Chr.2: 37119916-37119916 on Build GRCh38
rs773284307 Chr.2: 37120025-37120025 on Build GRCh38
rs746926144 Chr.2: 37119968-37119968 on Build GRCh38
rs747766352 Chr.2: 37120004-37120004 on Build GRCh38
rs979293852 Chr.2: 37120001-37120001 on Build GRCh38
rs772828788 Chr.2: 37120014-37120014 on Build GRCh38
rs780876414 Chr.2: 37119998-37119998 on Build GRCh38
rs144404132 Chr.2: 37120049-37120049 on Build GRCh38
rs765503875 Chr.2: 37120078-37120078 on Build GRCh38
rs966943599 Chr.2: 37119969-37119969 on Build GRCh38
rs957945467 Chr.2: 37119927-37119927 on Build GRCh38
rs776898858 Chr.2: 37120059-37120059 on Build GRCh38

View Full Product Details