Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99928904-99929153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJC3
Entrez Gene ID
349149
Gene Name
gap junction protein gamma 3
Gene Aliases
CX29, CX30.2, CX31.3, GJE1
UniGene
Hs.647524
Gene Chromosome Location
Chr.7: 99923269-99929620 on Build GRCh38
Transcripts
NM_181538.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6232625 c.510C>T p.R170R Chr.7: 99929111-99929111 on Build GRCh38
3267452 c.704C>T p.S235L Chr.7: 99928917-99928917 on Build GRCh38
6198932 c.697C>T p.L233L Chr.7: 99928924-99928924 on Build GRCh38
311462 c.585G>A p.M195I Chr.7: 99929036-99929036 on Build GRCh38
4550965 c.514G>A p.E172K Chr.7: 99929107-99929107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774027050 Chr.7: 99928969-99928969 on Build GRCh38
rs748522507 Chr.7: 99928935-99928935 on Build GRCh38
rs758559852 Chr.7: 99929109-99929109 on Build GRCh38
rs373506409 Chr.7: 99928983-99928983 on Build GRCh38
rs757817137 Chr.7: 99929068-99929068 on Build GRCh38
rs765434083 Chr.7: 99929055-99929055 on Build GRCh38
rs751656629 Chr.7: 99929111-99929111 on Build GRCh38
rs763838138 Chr.7: 99929071-99929071 on Build GRCh38
rs759448692 Chr.7: 99928974-99928974 on Build GRCh38
rs752804324 Chr.7: 99929096-99929096 on Build GRCh38
rs748551021 Chr.7: 99929130-99929130 on Build GRCh38
rs143497140 Chr.7: 99929023-99929023 on Build GRCh38
rs776612346 Chr.7: 99929063-99929064 on Build GRCh38
rs758650651 Chr.7: 99929013-99929013 on Build GRCh38
rs755928208 Chr.7: 99928993-99928993 on Build GRCh38
rs776998028 Chr.7: 99928933-99928933 on Build GRCh38
rs773731129 Chr.7: 99929039-99929039 on Build GRCh38
rs750596495 Chr.7: 99929003-99929003 on Build GRCh38
rs1026275866 Chr.7: 99929043-99929043 on Build GRCh38
rs778214490 Chr.7: 99929132-99929132 on Build GRCh38
rs990634745 Chr.7: 99929085-99929085 on Build GRCh38
rs770708878 Chr.7: 99929045-99929045 on Build GRCh38
rs73405465 Chr.7: 99929131-99929131 on Build GRCh38
rs755056506 Chr.7: 99929112-99929112 on Build GRCh38
rs111990958 Chr.7: 99929074-99929074 on Build GRCh38
rs1038562815 Chr.7: 99928959-99928959 on Build GRCh38
rs121908693 Chr.7: 99929052-99929052 on Build GRCh38
rs766648087 Chr.7: 99929110-99929110 on Build GRCh38
rs781712005 Chr.7: 99929120-99929120 on Build GRCh38
rs747288972 Chr.7: 99928964-99928964 on Build GRCh38
rs781659404 Chr.7: 99929070-99929070 on Build GRCh38
rs773042122 Chr.7: 99928949-99928949 on Build GRCh38
rs771020964 Chr.7: 99929055-99929056 on Build GRCh38
rs768493733 Chr.7: 99929095-99929096 on Build GRCh38
rs143828671 Chr.7: 99929024-99929024 on Build GRCh38
rs758651677 Chr.7: 99929123-99929123 on Build GRCh38
rs780043255 Chr.7: 99929018-99929018 on Build GRCh38
rs868504502 Chr.7: 99929094-99929095 on Build GRCh38
rs915119601 Chr.7: 99929094-99929094 on Build GRCh38
rs923827041 Chr.7: 99929117-99929117 on Build GRCh38
rs374090281 Chr.7: 99929032-99929032 on Build GRCh38
rs376850322 Chr.7: 99929021-99929021 on Build GRCh38
rs769486892 Chr.7: 99928940-99928940 on Build GRCh38
rs146848804 Chr.7: 99928928-99928928 on Build GRCh38
rs762906723 Chr.7: 99929089-99929090 on Build GRCh38
rs901784604 Chr.7: 99928956-99928956 on Build GRCh38
rs1025202730 Chr.7: 99929116-99929116 on Build GRCh38
rs1034450828 Chr.7: 99929067-99929067 on Build GRCh38
rs748905281 Chr.7: 99929040-99929040 on Build GRCh38
rs765935157 Chr.7: 99928958-99928958 on Build GRCh38
rs762525688 Chr.7: 99928957-99928957 on Build GRCh38
rs115480274 Chr.7: 99929051-99929051 on Build GRCh38
rs767367390 Chr.7: 99928980-99928980 on Build GRCh38

View Full Product Details