Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 11293114-11293339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT18
Entrez Gene ID
374378
Gene Name
polypeptide N-acetylgalactosaminyltransferase 18
Gene Aliases
GALNACT18, GALNT15, GALNTL4, GalNAc-T15, GalNAc-T18
UniGene
Hs.655152
Gene Chromosome Location
Chr.11: 11270874-11622045 on Build GRCh38
Transcripts
NM_198516.2 XM_006718224.2 XM_011520068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1627790 c.1513-8G>A p.? Chr.11: 11293201-11293201 on Build GRCh38
3443487 c.1548G>A p.V516V Chr.11: 11293158-11293158 on Build GRCh38
4686335 c.1537C>T p.Q513* Chr.11: 11293169-11293169 on Build GRCh38
1317395 c.1546G>A p.V516M Chr.11: 11293160-11293160 on Build GRCh38
4017882 c.1516G>A p.V506M Chr.11: 11293190-11293190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762703454 Chr.11: 11293211-11293211 on Build GRCh38
rs201537431 Chr.11: 11293230-11293230 on Build GRCh38
rs1051732897 Chr.11: 11293280-11293280 on Build GRCh38
rs1025724833 Chr.11: 11293282-11293282 on Build GRCh38
rs112108154 Chr.11: 11293241-11293241 on Build GRCh38
rs898581004 Chr.11: 11293262-11293262 on Build GRCh38
rs755705416 Chr.11: 11293211-11293212 on Build GRCh38
rs766192930 Chr.11: 11293242-11293242 on Build GRCh38
rs548962000 Chr.11: 11293140-11293140 on Build GRCh38
rs529247669 Chr.11: 11293139-11293139 on Build GRCh38
rs754105385 Chr.11: 11293183-11293183 on Build GRCh38
rs771187705 Chr.11: 11293215-11293215 on Build GRCh38
rs1045614537 Chr.11: 11293259-11293259 on Build GRCh38
rs1034511859 Chr.11: 11293305-11293305 on Build GRCh38
rs751676341 Chr.11: 11293167-11293167 on Build GRCh38
rs759676464 Chr.11: 11293218-11293218 on Build GRCh38
rs779824658 Chr.11: 11293190-11293190 on Build GRCh38
rs368789058 Chr.11: 11293202-11293202 on Build GRCh38
rs768067191 Chr.11: 11293177-11293177 on Build GRCh38
rs185402461 Chr.11: 11293219-11293219 on Build GRCh38
rs773089871 Chr.11: 11293207-11293207 on Build GRCh38
rs374763409 Chr.11: 11293233-11293233 on Build GRCh38
rs386750685 Chr.11: 11293252-11293253 on Build GRCh38
rs769569442 Chr.11: 11293205-11293205 on Build GRCh38
rs925759620 Chr.11: 11293251-11293251 on Build GRCh38
rs369723200 Chr.11: 11293243-11293243 on Build GRCh38
rs774539122 Chr.11: 11293217-11293217 on Build GRCh38
rs752665485 Chr.11: 11293223-11293223 on Build GRCh38
rs113829057 Chr.11: 11293275-11293275 on Build GRCh38
rs753239923 Chr.11: 11293179-11293179 on Build GRCh38
rs1001743636 Chr.11: 11293299-11293299 on Build GRCh38
rs138326637 Chr.11: 11293252-11293252 on Build GRCh38
rs755750886 Chr.11: 11293308-11293308 on Build GRCh38
rs372092468 Chr.11: 11293180-11293180 on Build GRCh38
rs1037938768 Chr.11: 11293290-11293291 on Build GRCh38
rs376700751 Chr.11: 11293201-11293201 on Build GRCh38
rs887143673 Chr.11: 11293294-11293294 on Build GRCh38
rs139933398 Chr.11: 11293169-11293169 on Build GRCh38
rs150020084 Chr.11: 11293188-11293188 on Build GRCh38
rs754270512 Chr.11: 11293240-11293240 on Build GRCh38
rs1008213568 Chr.11: 11293298-11293298 on Build GRCh38
rs886452088 Chr.11: 11293286-11293286 on Build GRCh38
rs763258008 Chr.11: 11293143-11293143 on Build GRCh38
rs992545379 Chr.11: 11293212-11293212 on Build GRCh38
rs1005067086 Chr.11: 11293287-11293287 on Build GRCh38
rs568876193 Chr.11: 11293155-11293155 on Build GRCh38
rs959777482 Chr.11: 11293307-11293307 on Build GRCh38
rs750010154 Chr.11: 11293210-11293210 on Build GRCh38
rs551200358 Chr.11: 11293191-11293191 on Build GRCh38
rs149641477 Chr.11: 11293253-11293253 on Build GRCh38
rs772824227 Chr.11: 11293141-11293141 on Build GRCh38
rs561697544 Chr.11: 11293309-11293309 on Build GRCh38

View Full Product Details