Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33747128-33747401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO3
Entrez Gene ID
26273
Gene Name
F-box protein 3
Gene Aliases
FBA, FBX3
UniGene
Hs.406787
Gene Chromosome Location
Chr.11: 33740937-33774531 on Build GRCh38
Transcripts
NM_012175.3 NM_033406.2 XM_011519980.1 XM_011519981.2 XM_011519982.1 XM_017017533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6209205 c.1156T>C p.Y386H Chr.11: 33747213-33747213 on Build GRCh38
2115281 c.1132G>C p.G378R Chr.11: 33747237-33747237 on Build GRCh38
6132477 c.1063A>T p.I355F Chr.11: 33747306-33747306 on Build GRCh38
926554 c.1173C>A p.I391I Chr.11: 33747196-33747196 on Build GRCh38
4461923 c.1226C>T p.S409F Chr.11: 33747143-33747143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761061269 Chr.11: 33747348-33747348 on Build GRCh38
rs1038107723 Chr.11: 33747155-33747155 on Build GRCh38
rs922004916 Chr.11: 33747169-33747169 on Build GRCh38
rs752650063 Chr.11: 33747298-33747298 on Build GRCh38
rs1051335599 Chr.11: 33747245-33747245 on Build GRCh38
rs201921995 Chr.11: 33747255-33747255 on Build GRCh38
rs1018210617 Chr.11: 33747291-33747291 on Build GRCh38
rs780005389 Chr.11: 33747293-33747293 on Build GRCh38
rs774447321 Chr.11: 33747337-33747337 on Build GRCh38
rs757065450 Chr.11: 33747149-33747149 on Build GRCh38
rs778826011 Chr.11: 33747156-33747156 on Build GRCh38
rs267602863 Chr.11: 33747223-33747223 on Build GRCh38
rs375297104 Chr.11: 33747362-33747362 on Build GRCh38
rs776013635 Chr.11: 33747347-33747347 on Build GRCh38
rs141615775 Chr.11: 33747199-33747199 on Build GRCh38
rs777801349 Chr.11: 33747309-33747309 on Build GRCh38
rs768286240 Chr.11: 33747176-33747176 on Build GRCh38
rs146257723 Chr.11: 33747163-33747163 on Build GRCh38
rs772353179 Chr.11: 33747342-33747342 on Build GRCh38
rs139508675 Chr.11: 33747246-33747246 on Build GRCh38
rs148483444 Chr.11: 33747170-33747170 on Build GRCh38
rs776273163 Chr.11: 33747371-33747371 on Build GRCh38
rs144350497 Chr.11: 33747247-33747247 on Build GRCh38
rs911907549 Chr.11: 33747261-33747261 on Build GRCh38
rs748196567 Chr.11: 33747186-33747186 on Build GRCh38
rs557360966 Chr.11: 33747338-33747338 on Build GRCh38
rs370192164 Chr.11: 33747217-33747217 on Build GRCh38
rs753149454 Chr.11: 33747235-33747235 on Build GRCh38
rs771128018 Chr.11: 33747336-33747336 on Build GRCh38
rs368515184 Chr.11: 33747227-33747227 on Build GRCh38
rs903846856 Chr.11: 33747329-33747329 on Build GRCh38
rs267602862 Chr.11: 33747172-33747172 on Build GRCh38
rs201350873 Chr.11: 33747256-33747256 on Build GRCh38
rs147718954 Chr.11: 33747294-33747294 on Build GRCh38
rs768008825 Chr.11: 33747229-33747229 on Build GRCh38
rs750177328 Chr.11: 33747289-33747289 on Build GRCh38
rs774694608 Chr.11: 33747226-33747226 on Build GRCh38
rs749438310 Chr.11: 33747323-33747323 on Build GRCh38
rs763139751 Chr.11: 33747196-33747196 on Build GRCh38
rs568578146 Chr.11: 33747279-33747279 on Build GRCh38
rs758134989 Chr.11: 33747292-33747292 on Build GRCh38
rs200315129 Chr.11: 33747233-33747233 on Build GRCh38
rs769799155 Chr.11: 33747190-33747190 on Build GRCh38
rs537531760 Chr.11: 33747283-33747283 on Build GRCh38
rs756937035 Chr.11: 33747280-33747280 on Build GRCh38

View Full Product Details